Skip to Content
Merck
All Photos(2)

Key Documents

850335C

Avanti

12:0 PC (DLPC)

1,2-dilauroyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-didodecanoyl-sn-glycero-3-phosphocholine; DLPC; PC(12:0/12:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H64NO8P
CAS Number:
Molecular Weight:
621.83
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850335C-25mg)
pkg of 1 × 20 mL (850335C-500mg)
pkg of 2 × 4 mL (850335C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850335C

concentration

10 mg/mL (850335C-25mg)
25 mg/mL (850335C-200mg)
25 mg/mL (850335C-500mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@@H](OC(=O)CCCCCCCCCCC)COC(=O)CCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C32H64NO8P/c1-6-8-10-12-14-16-18-20-22-24-31(34)38-28-30(29-40-42(36,37)39-27-26-33(3,4)5)41-32(35)25-23-21-19-17-15-13-11-9-7-2/h30H,6-29H2,1-5H3/t30-/m0/s1

InChI key

IJFVSSZAOYLHEE-PMERELPUSA-N

General description

The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.
The mucosal layer of the colon has phosphatidylcholine (PC). PCs carry fatty acids in the sn-1 and sn-2 positions, that are covalently attached to a glycerol through ester bonds.

Application

12:0 PC (DLPC/1,2-dilauroyl-sn-glycero-3-phosphocholine) may be used as a standard to quantify phosphatidylcholine. It has also been used as a lipid standard in the optimization of the liquid chromatography−mass spectrometry (LC-MS) method conditions.

Biochem/physiol Actions

Phosphatidylcholine (PC) has the ability to block the penetrance of bacteria by acting as a surfactant.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850335C-500mg)
5 mL Clear Glass Sealed Ampule (850335C-200mg)
5 mL Clear Glass Sealed Ampule (850335C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Extraction and profiling of plant polar glycerol lipids
Liu Y and Wang X
Bio-protocol, 6, 1849e-1849e null
Comprehensive untargeted lipidomic analysis using core
Narvaaez R, et al.
Journal of Chromatography A, 1440, 123-134 null
Nhiem Tran et al.
The journal of physical chemistry letters, 9(12), 3397-3402 (2018-05-29)
The transition between the lyotropic liquid crystalline lamellar and the bicontinuous cubic mesophase drives multiple fundamental cellular processes involving changes in cell membrane topology, including endocytosis and membrane budding. While several theoretical models have been proposed to explain this dynamic
John P Kowalski et al.
Chemical research in toxicology, 32(12), 2488-2498 (2019-12-05)
Cytochrome P450 4B1 (CYP4B1) has been explored as a candidate enzyme in suicide gene systems for its ability to bioactivate the natural product 4-ipomeanol (IPO) to a reactive species that causes cytotoxicity. However, metabolic limitations of IPO necessitate discovery of
Inflammatory Bowel Disease
Parian AM, et al.
Integrative Medicine, 501-516 null

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service