Skip to Content
Merck
All Photos(2)

Key Documents

810280P

Avanti

TopFluor Lyso PA

Avanti Research - A Croda Brand 810280P, powder

Synonym(s):

1-{12-[4-(dipyrrometheneboron difluoride)butanoyl]amino}dodecanoyl-2-hydroxy-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54BF2N4O8P
CAS Number:
Molecular Weight:
702.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (810280P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810280P

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidic acid (LPA) is a water-soluble, bioactive phospholipid. It is seen in mammalian cells and tissues, including blood. LPA is usually formed during the synthesis of phospholipid of cell membranes. It is produced by various cells like, activated platelets, epithelial cells, leukocytes, neuronal cells and tumor cells.

Application

TopFluor Lyso PA may be used as the chromophore and as a component in the formation of the Langmuir-Blodgett (LB) film.

Biochem/physiol Actions

Lysophosphatidic acid (LPA) is very essential in signaling. It may alter proliferation, survival, cytoskeletal changes and calcium influx. It plays a key role in neointimal formation, endothelium dysfunction, monocyte attraction and adhesion.

Packaging

5 mL Amber Glass Screw Cap Vial (810280P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidic acid triggers apoptosis in HeLa cells through the upregulation of tumor necrosis factor receptor superfamily member 21
Dong Y, et al.
Mediators of Inflammation (2017)
Lysophosphatidic Acid Triggers Apoptosis in HeLa Cells through the Upregulation of Tumor Necrosis Factor Receptor Superfamily Member 21
Dong Y, et al.
Mediators of Inflammation (2017)
Diffusional motion as a gauge of fluidity and interfacial adhesion. Supported alkylphosphonate monolayers
Baumler SM, et al.
Journal of Colloid and Interface Science, 468, 145-155 (2016)
Lysophosphatidic acid (LPA) receptors: signaling properties and disease relevance
Lin ME, et al.
Prostaglandins & other lipid mediators, 91(3-4), 130-138 (2010)
Lauren P Saunders et al.
The Journal of biological chemistry, 286(34), 30130-30141 (2011-07-02)
Autotaxin (ATX) is a secreted lysophospholipase D that hydrolyzes lysophosphatidylcholine (LPC) into lysophosphatidic acid (LPA), initiating signaling cascades leading to cancer metastasis, wound healing, and angiogenesis. Knowledge of the pathway and kinetics of LPA synthesis by ATX is critical for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service