Skip to Content
Merck
All Photos(5)

Key Documents

VT000122

Millipore

Escherichia coli WDCM 00012 Vitroids

greener alternative

15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

E. coli WDCM 00012 Vitroids, Escherichia coli WDCM 00012 Vitroids, Certified reference material, Microbiological standards, E. coli WDCM 00012 Vitroids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

ISO 17034
ISO/IEC 17025

form

disc (hydrosoluble)
solid

shelf life

limited shelf life, expiry date on the label

packaging

pack of 10

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

sustainability

Greener Alternative Product

application(s)

environmental
food and beverages

microbiology

greener alternative category

storage temp.

−20°C

suitability

Escherichia coli
Escherichia spp.
coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Escherichia coli WDCM 00012 Vitroids are Certified Microorganisms Standards (Certified Microbiological Reference Materials) which can be used as quantitative and qualitative control organisms. They contain colony forming units (CFUs) in the range of 15-80 CFU per disc. The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms. These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need to maintain control strains in the test laboratory and guarantee an authenticated control culture for every quality control test. Vitroids are derived from a traceable culture obtained freeze-dried from CECT®.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of our four categories of greener alternatives. Click here to view its DfS scorecard.

Application

Escherichia coli WDCM 00012 Vitroids are used as reference strains in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for use in research studies and educational purposes.

Features and Benefits

  • Ready-to-use at convenient CFU ranges
  • No dilution required
  • Quick rehydration in 10 minutes
  • 2-3 years of shelf life in a standard -20 °C freezer
  • Complete with a comprehensive certificate of analysis
  • No maintenance of stock cultures
  • No recovery time or pre-enrichment step is necessary
  • Robust, efficient, and reliable
  • Easy-to-use
  • Cost-effective

Other Notes

Other cell culture collections:
  • ATCC® 8739
  • NCTC 12923
  • DSM 1576
  • LMG 8063
  • CIP 53.126
Click here to access the BacDive database for more info on metabolism, genome, growth media, and more
Origin Strain No. CECT® 516

Strain History: < NCIMB, 1953 < ATCC < I.C. Gunsalus < G.C. Crooks

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Related Content

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service