Skip to Content
Merck
All Photos(2)

Key Documents

810195P

Avanti

18:1-12:0 NBD PS

Avanti Research - A Croda Brand 810195P, powder

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoserine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H73N6O13P
CAS Number:
Molecular Weight:
901.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810195P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810195P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylserine (PS) is an acidic membrane phospholipid present in human cerebral cortex. It is majorly present in the inner leaflet of plasma membrane. NBD PS contains the fluorophore, 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) labelled at the headgroup of PS.

Application

18:1-12:0 NBD PS or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoserine (ammonium salt), may be used to measure the phosphatidylserine decarboxylase (Psd1) activity.

Biochem/physiol Actions

Phosphatidylserine (PS) is involved in maintaining the nerve cell membrane and myelin sheath. Phosphatidylserine (PS) is associated with Akt, protein kinase C (PKC) and Raf-1 signaling.

Packaging

5 mL Amber Glass Screw Cap Vial (810195P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylserine and the human brain
Glade MJ and Smith K
Nutrition null
Lipid Homeostasis Is Maintained by Dual Targeting of the Mitochondrial PE Biosynthesis Enzyme to the ER
Jonathan R, et al.
Developmental Cell null
Structural Basis for Phosphatidylethanolamine Biosynthesis by Bacterial Phosphatidylserine Decarboxylase
Yasunori W, et al.
Structure null
Organization and dynamics of N-(7-nitrobenz-2-oxa-1, 3-diazol-4-yl)-labeled lipids: a fluorescence approach
Mukherjee S, et al.
Chemistry and Physics of Lipids null
Jonathan R Friedman et al.
Developmental cell, 44(2), 261-270 (2018-01-02)
Spatial organization of phospholipid synthesis in eukaryotes is critical for cellular homeostasis. The synthesis of phosphatidylcholine (PC), the most abundant cellular phospholipid, occurs redundantly via the ER-localized Kennedy pathway and a pathway that traverses the ER and mitochondria via membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service