Skip to Content
Merck
All Photos(2)

Key Documents

810157P

Avanti

14:0 Liss Rhod PE

Avanti Research - A Croda Brand 810157P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H97N4O14PS2
CAS Number:
Molecular Weight:
1193.53
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810157P-1mg)
pkg of 5 × 1 mg (810157P-5mg)
pkg of 5 × 2 mg (810157P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810157P

shipped in

dry ice

storage temp.

−20°C

Application

14:0 Liss Rhod PE or 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(lissamine rhodamine B sulfonyl) (ammonium salt) has been used in the preparation of fluorescent labeled vesicles to understand lipid fouling behaviour of glycosphingolipid and lipopolysaccharide on nanofiltration membranes. 14:0 Liss Rhod PE may also be used: in lipid mixing assay to prepare labeled phospholipid vesicle, for the formation of vesicles in surface plasmon resonance (SPR)-binding experiments and in the preparation of red labeled liposome

Packaging

5 mL Amber Glass Screw Cap Vial (810157P-10mg)
5 mL Amber Glass Screw Cap Vial (810157P-1mg)
5 mL Amber Glass Screw Cap Vial (810157P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro Membrane Interaction and Liposome Fusion Assays Using Recombinant Hepatitis C Virus Envelope Protein E2
Yelamos B, et al.
Bio-protocol, 8(23), 43-47 (2018)
A Natural Human IgM that Binds to Gangliosides Is Therapeutic in Murine Models of Amyotrophic Lateral Sclerosis
Warrington AE and ME Jonah Shaver
Disease models & mechanisms, 8(23), 43-47 (2018)
Glycosphingolipids enhance bacterial attachment and fouling of nanofiltration membranes
Haas R, et al.
Environmental Sciences, 2(2), 43-47 (2015)
Induction of differential T-cell epitope by plain-and liposome-coupled antigen
Taneichi M, et al.
Bioconjugate Chemistry, 8(23), 43-47 (2018)
[A leader for health care and service].
S Stubbendorff
Vardfacket, 13(19), 3-3 (1989-11-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service