Skip to Content
Merck
All Photos(1)

Key Documents

800100O

Avanti

18:1-2:0 DG

1-oleoyl-2-acetyl-sn-glycerol, neat oil

Synonym(s):

1--(9Z-octadecenoyl)-2-acetoyl-sn-glycerol; DG(18:1(9Z)/2:0/0:0); OAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H42O5
CAS Number:
Molecular Weight:
398.58
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 10 mg (with screw cap (800100O-10mg))
pkg of 1 × 25 mg (with screw cap (800100O-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand 800100O

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCC\C=C/CCCCCCCC)CO)C(=O)C

InChI

1S/C23H42O5/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)27-20-22(19-24)28-21(2)25/h10-11,22,24H,3-9,12-20H2,1-2H3/b11-10-/t22-/m0/s1

InChI key

PWTCCMJTPHCGMS-YRBAHSOBSA-N

General description

18:1-2:0 DG, also called 1-oleoyl-2-acetyl-sn-glycerol (OADG), is a membrane permeable and synthetic analog of diacyl glycerol (DAG).

Application

18:1-2:0 DG (1-oleoyl-2-acetyl-sn-glycerol) may be used:
  • in the induction of superoxide generation in neutrophils
  • as a substrate for platelet-activating factor acetyl hydrolases
  • as a protein kinase C (PKC) activator in adrenal glands

Biochem/physiol Actions

18:1-2:0 DG (OAG) halts the inhibition of glucose uptake by the phospholipase C (PLC) inhibitor, U73122. It also inhibits mitochondrial permeability transition pore (MPTP) formation and prevents fatty acid release and necrosis in lung fibroblasts. OAG is a protein kinase C (PKC) activator. It also inhibits TWIK-related acid-sensitive (TASK)-like potassium channels in peripheral arterial chemoreceptors. OAG stimulates 5-lipooxygenase (5-LO) in human polymorphonuclear leukocytes (PMNL).

Packaging

5 mL PTFE Vial with Screw Cap (800100O-10mg)
5 mL PTFE Vial with Screw Cap (800100O-25mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells: Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration: Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

sn-1,2-dioctanoylglycerol: a Cell Permeable Diacylglycerol That Mimics Phorbol Diester Action on the Epidermal Growth Factor Receptor and Mitogenesis.
Davis RJ, et al.
The Journal of Biological Chemistry, 260, 1562-1566 (1985)
J H Min et al.
Biochemistry, 38(39), 12935-12942 (1999-10-03)
Human plasma platelet activating factor acetylhydrolase (pPAF-AH) is a phospholipase A(2) that specifically hydrolyzes the sn-2 ester of platelet activating factor (PAF) and of phospholipids with oxidatively truncated sn-2 fatty acyl chains. pPAF-AH is bound to lipoproteins in vivo, and
Exogenous sn-1,2-diacylglycerols Containing Saturated Fatty Acids Functions as Bioregulators of Protein Kinase C in Human Platelets.
Lapetina EG, et al.
The Journal of Biological Chemistry, 260, 1358-1361 (1985)
Pawan Bista et al.
The Journal of physiology, 593(1), 127-144 (2015-01-06)
During the behavioural states of sleep and wakefulness thalamocortical relay neurons fire action potentials in high frequency bursts or tonic sequences, respectively. The modulation of specific K(+) channel types, termed TASK and TREK, allows these neurons to switch between the
Diacylglycerols and Phorbol Diesters Induce Leukemic Cell Differentiation via a Common Mechanism.
Ebeling JG, et al.
Proceedings of the National Academy of Sciences of the USA, 82, 815-819 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service