Skip to Content
Merck
All Photos(1)

Documents

790304C

Avanti

Coag Reagent I

Avanti Research - A Croda Brand 790304C

Synonym(s):

Coag Reagent, Coagulation Reagent I, contains DOPE:DOPS:DOPC (5:3:2 w/w)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (790304C-25mg)
pkg of 2 × 4 mL (790304C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790304C

concentration

10 mg/mL (790304C-25mg)
25 mg/mL (790304C-200mg)

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Coag. Reagent I has been used for the preparation of small unilamellar vesicles (SUVs).

Packaging

5 mL Clear Glass Sealed Ampule (790304C-200mg)
5 mL Clear Glass Sealed Ampule (790304C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M van den Besselaar et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 4(6), 895-903 (1993-12-01)
The activated partial thromboplastin time (aPTT) is the most popular test for monitoring of heparin therapy. The purpose of the present study was to show that an aPTT reagent with good response to heparin can be prepared from synthetic phosphoglycerides.
Nuclear magnetic resonance observation of ?-synuclein membrane interaction by monitoring the acetylation reactivity of its lysine side chains
Lee JH, et al.
Biochemistry, 55(35), 4949-4959 (2016)
Three-Dimensional Heterogeneous Structure Formation on a Supported Lipid Bilayer Disclosed by Single-Particle Tracking
Zhong Y & Wang G
Langmuir, 34(39), 11857-11865 (2018)
A M van den Besselaar et al.
Clinical chemistry, 43(7), 1215-1222 (1997-07-01)
To contribute to the development of a reference reagent for monitoring heparin therapy, a lyophilized partial thromboplastin time (PTT) reagent was prepared from synthetic dioleoylphosphatidylcholine, dioleoylphosphatidylserine, and dioleoylphosphatidylethanolamine, with colloidal silica as activator. The reagent, coded 91/558, was contained in
Yaning Zhong et al.
Langmuir : the ACS journal of surfaces and colloids, 34(39), 11857-11865 (2018-09-02)
Three-dimensional (3D) single-particle tracking was employed to study the lipid membrane morphology change at different pHs on glass supported lipid bilayers (SLBs) [1,2-dioleoyl- sn-glycero-3-phosphoethanolamine/1,2-dioleoyl- sn-glycero-3-phospho-l-serine (sodium salt)/1,2-dioleoyl- sn-glycero-3-phosphocholine = 5:3:2]. Fluorescently tagged, carboxylated polystyrene nanoparticles (of 100 nm) were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service