Key Documents
Z265942
Sigma-Aldrich
Solid-glass beads
soda-lime glass, diam. 5 mm
About This Item
UNSPSC Code:
41102422
NACRES:
NB.43
material
soda-lime glass
packaging
pack of 1 ea
diam.
5 mm
General description
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Choose from one of the most recent versions:
Certificates of Analysis (COA)