Skip to Content
Merck

1.04016

Glass beads 4 mm

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NB.51
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

storage temp.

no temp limit

Quality Level

General description

Glass beads can be used to purify, enrich or separate samples of solids, liquids or gases. They offer excellent batch consistency to ensure reliable, reproducible results.

Application

Glass beads 4 mm can be used as a supporting layer in a fluidized bed reactor. It is also used in shaken glass cultures to promote quick bacterial growth and reduce the formation of aggregates.

Analysis Note

Identity: conforms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fouling mitigation in Anaerobic Membrane Bioreactor using fluidized glass beads: Evaluation fitness for purpose of ceramic membranes
Duppenbecker B, et al.
Journal of Membrane Science , 537, 69-82 (2017)
Growth characteristics and morphological changes of fish pathogen Nocardia seriolae in submerged culture
Shimahara Y, et al.
Aquaculture Science, 58(3), 429-431 (2010)

Global Trade Item Number

SKUGTIN
104016050004022536669519

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service