Z146226
Rectangular TLC developing tanks, complete
L × H × W 12.1 cm × 10.8 cm × 8.3 cm
Synonym(s):
TLC Developing Chambers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
Recommended Products
technique(s)
thin layer chromatography (TLC): suitable
L × H × W
12.1 cm × 10.8 cm × 8.3 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results.
Application
Aldrich® rectangular TLC developing tanks may be used to develop TLC plates.
Footnote
TLC Developing Tanks come complete with the developing tank and lid
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
An overview on thin layer chromatography
Bele, Archana A., and Anubha Khale.
International Journal of Pharmaceutical Sciences and Research, 2, 256-267 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service