Skip to Content
Merck
All Photos(1)

Documents

W400201

Sigma-Aldrich

Methyl methacrylate

99%, stabilized

Synonym(s):

Methyl 2-methylacrylate, Methyl 2-methylprop-2-enoate, Methyl 2-methylpropenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH3
CAS Number:
Molecular Weight:
100.12
FEMA Number:
4002
Beilstein:
605459
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.647
NACRES:
NA.21

biological source

synthetic

Quality Level

vapor density

3.5 (vs air)

vapor pressure

29 mmHg ( 20 °C)

Assay

99%

autoignition temp.

815 °F

contains

hydroquinone monomethyl ether as stabilizer

expl. lim.

12.5 %

refractive index

n20/D 1.414 (lit.)

bp

100 °C (lit.)

mp

−48 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity

SMILES string

COC(=O)C(C)=C

InChI

1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3

InChI key

VVQNEPGJFQJSBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl methacrylate is mainly used in the preparation of polymethyl methacrylate, a bone cement material.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational Hazards in the Health Professions, 142-142 (1989)
Kevin N Johnson et al.
The Annals of thoracic surgery, 99(6), 1936-1943 (2015-04-30)
Successful repair of recurrent pectus excavatum (PE) after failed open procedure has been reported using minimally invasive repair (MIRPE) and open approaches. Neither approach alone may be adequate for some patients. A hybrid technique for repair is presented for revision
Terumasa Kato et al.
The Journal of organic chemistry, 78(17), 8739-8747 (2013-08-15)
We and others have previously reported the intermolecular umpolung reactions of Michael acceptors catalyzed by an N-heterocyclic carbene (NHC). The representative tail-to-tail dimerization of methyl methacrylate (MMA) has now been intensively investigated, leading to the following conclusions: (1) The catalysis
Alina Badina et al.
Clinical orthopaedics and related research, 471(11), 3701-3705 (2013-05-16)
Acetabular fractures are rare in children and can be complicated by premature fusion of the triradiate cartilage resulting in secondary acetabular dysplasia. Early recognition and treatment of a physeal bar in this location can be difficult. The purpose of this
Fabrice Audouin et al.
Biomacromolecules, 13(11), 3787-3794 (2012-10-20)
Amino-functional macroporous monoliths from polymerized high internal phase emulsion (polyHIPE) were surface modified with initiators for atom transfer radical polymerization (ATRP). The ATRP initiator groups on the polyHIPE surface were successfully used to initiate activator regeneration by electron transfer (ARGET)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service