Skip to Content
Merck
All Photos(3)

Key Documents

445746

Sigma-Aldrich

Poly(methyl methacrylate)

average Mw ~350,000 by GPC

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

autoignition temp.

580 °F

mol wt

average Mw ~350,000 by GPC

reduced viscosity

150-160 cm3/g(20 °C)(lit.)

acid number

<1 mg KOH/g

transition temp

Tg (DSC) 105 °C (midpoint)

solubility

alcohols and aliphatic hydrocarbons: insoluble
esters, ketones, aromatics and glycol ethers: soluble

density

1.17 g/mL at 25 °C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The thermal degradation behaviour of PMMA ( avg Mol. wt. ~350,000 g mol-1)was investigated in inert atmosphere, four weight loss steps were observed.

Application

PMMA (avg. Mol wt. ~350,000 g mol-1) containing Fe2O3 and TiO2 fillers have possible uses as flame retardants.
Pigment-wetting binder for coatings.

Features and Benefits

Contributes hardness, weather resistance, light fastness and resistance to chemicals.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal degradation of poly (methyl methacrylate)(PMMA): modelling of DTG and TG curves.
Ferriol M., et al.
Polymer Degradation and Stability, 79(2), 271-281 (2003)
Influence of TiO2 and Fe2O3 fillers on the thermal properties of poly (methyl methacrylate)(PMMA).
Laachachi A.,et al.
Material Matters, 59(1), 36-39 (2005)
Distortions of iris and anterior lens capsule OCT images by ICRS.
Hosik Hwang et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(3), 155-156 (2013-03-01)
Bo Chen et al.
Journal of controlled release : official journal of the Controlled Release Society, 140(3), 203-209 (2009-05-26)
Water-soluble polymers for the delivery of chemotherapeutic drugs passively target solid tumors as a consequence of reduced renal clearance and the enhanced permeation and retention (EPR) effect. Elimination of the polymers in the kidney occurs due to filtration through biological
Clément M L Werner et al.
The Journal of bone and joint surgery. American volume, 95(7), 577-584 (2013-04-05)
In the treatment of vertebral compression fractures, vertebral body stenting with an expandable scaffold inserted before application of the bone cement was developed to impede secondary loss of vertebral height encountered in patients treated with balloon kyphoplasty. The purpose of

Articles

The detection and quantification of biomarkers are essential for medical diagnostics, environmental monitoring, and bioresearch.

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service