Skip to Content
Merck
All Photos(1)

Key Documents

W286400

Sigma-Aldrich

Phenethyl formate

≥96%, FG

Synonym(s):

Benzyl carbinyl formate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH2CH2C6H5
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2864
Council of Europe no.:
350
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.083
NACRES:
NA.21
Organoleptic:
green; herbaceous; rose
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥96%

refractive index

n20/D 1.5075 (lit.)

bp

226 °C (lit.)

density

1.058 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; herbaceous; rose

storage temp.

2-8°C

SMILES string

[H]C(=O)OCCc1ccccc1

InChI

1S/C9H10O2/c10-8-11-7-6-9-4-2-1-3-5-9/h1-5,8H,6-7H2

InChI key

IKDIJXDZEYHZSD-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fangfang Zeng et al.
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service