Skip to Content
Merck
All Photos(1)

Key Documents

W280011

Sigma-Aldrich

1-Octanol

natural, ≥98%, FCC

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
FEMA Number:
2800
Beilstein:
1697461
EC Number:
Council of Europe no.:
54c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.006
NACRES:
NA.21
grade:
Kosher
natural
food allergen:
no known allergens

grade

Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 172.515

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥98%

form

liquid

autoignition temp.

523 °F

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; citrus; waxy; floral; sweet

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Southwell et al.
The Journal of investigative dermatology, 80(6), 507-514 (1983-06-01)
Penetration-enhancing solvents may modify diffusional routes through the stratum corneum and the partitioning of drugs into these pathways. To investigate the mechanisms of action of enhancers, steady-state fluxes were measured in vitro for polar methanol, nonpolar octanol, and an intermediate
John A Arnott et al.
Expert opinion on drug discovery, 7(10), 863-875 (2012-09-21)
The role of lipophilicity in drug discovery and design is a critical one. Lipophilicity is a key physicochemical property that plays a crucial role in determining ADMET (absorption, distribution, metabolism, excretion, and toxicity) properties and the overall suitability of drug
B D Anderson et al.
The Journal of investigative dermatology, 93(2), 280-286 (1989-08-01)
The permeability coefficients (kp) of a series of methyl-substituted p-cresols were determined in human stratum corneum along with their partition coefficients (PC) between water and untreated stratum corneum, delipidized stratum corneum, octanol, and heptane. The PC values were identical in
Cristian Gobbo et al.
ACS nano, 6(12), 10684-10698 (2012-12-12)
Controlling the molecular conformation of oligomers on surfaces through noncovalent interactions symbolizes an important approach in the bottom-up patterning of surfaces with nanoscale precision. Here we report on the design, synthesis, and scanning tunneling microscopy (STM) investigation of linear oligoamides
R O Potts et al.
Pharmaceutical research, 9(5), 663-669 (1992-05-01)
Published permeability coefficient (Kp) data for the transport of a large group of compounds through mammalian epidermis were analyzed by a simple model based upon permeant size [molecular volume (MV) or molecular weight (MW)] and octanol/water partition coefficient (Koct). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service