Skip to Content
Merck
All Photos(1)

Key Documents

W243701

Sigma-Aldrich

Ethyl heptanoate

≥98%, FG

Synonym(s):

Ethyl enanthate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COOC2H5
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2437
Beilstein:
1752311
EC Number:
Council of Europe no.:
365
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39022453
PubChem Substance ID:
Flavis number:
9.093
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.412 (lit.)

bp

188-189 °C (lit.)

mp

−66 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; pineapple; rum

SMILES string

CCCCCCC(=O)OCC

InChI

1S/C9H18O2/c1-3-5-6-7-8-9(10)11-4-2/h3-8H2,1-2H3

InChI key

TVQGDYNRXLTQAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl heptanoate is one of the key flavor volatiles identified in Luzhou-flavor raw spirits and gingko wine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Volatile compounds of raw spirits from different distilling stages of Luzhou-flavor spirit
Zheng, Jia, et al
Food Science and Technology Research, 20(2), 283-293 (2014)
J Enrique Cometto-Muñiz et al.
Experimental brain research, 158(2), 196-206 (2004-04-28)
A series of experiments probed into the degree of chemosensory detection additivity exhibited by mixtures of ethyl propanoate and heptanoate in terms of their trigeminal detectability via nasal pungency (i.e., irritation) and eye irritation. Nasal pungency was tested in subjects
Mohammad Rafienia et al.
Current drug delivery, 6(2), 184-191 (2009-05-20)
In situ forming biodegradable polymeric systems were prepared from Poly (DL-lactide-co-glycolide), RG504H (50:50, lactide:glycolide), RG756 (75:25) and mixture of them. They were dissolved in N-methyl-2-pyrrolidone (33% w/w) and mixed with betamethasone acetate (BTMA, 5 and 10% w/w) and ethyl heptanoate
Monographs on fragrance raw materials.
D L Opdyke
Food and cosmetics toxicology, 19(2), 237-254 (1981-04-01)
J Enrique Cometto-Muñiz et al.
Behavioural brain research, 156(1), 115-123 (2004-10-12)
The investigation explored the olfactory detectability of two chemically and structurally similar esters, ethyl propanoate and ethyl heptanoate, presented singly and in mixtures. Initially, we measured concentration-detection (i.e., psychometric) functions for the odor of ethyl propanoate and ethyl heptanoate presented

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service