Skip to Content
Merck
All Photos(3)

Documents

112364

Sigma-Aldrich

Ethyl heptanoate

ReagentPlus®, 99%

Synonym(s):

Ethyl enanthate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COOC2H5
CAS Number:
Molecular Weight:
158.24
Beilstein:
1752311
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022453
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.412 (lit.)

bp

188-189 °C (lit.)

mp

−66 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(=O)OCC

InChI

1S/C9H18O2/c1-3-5-6-7-8-9(10)11-4-2/h3-8H2,1-2H3

InChI key

TVQGDYNRXLTQAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl heptanoate, an aroma compound, was released from a series of sodium caseinate-stabilized, n-eicosane emulsions during the investigation of solid and liquid lipid droplet concentration.

Application

Ethyl heptanoate was used in a nickel nanoparticles-catalysed, transfer hydrogenation of olefins.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transfer hydrogenation of olefins catalysed by nickel nanoparticles.
Alonso F, et al.
Tetrahedron, 65(51), 10637-10643 (2009)
Mohammad Rafienia et al.
Current drug delivery, 6(2), 184-191 (2009-05-20)
In situ forming biodegradable polymeric systems were prepared from Poly (DL-lactide-co-glycolide), RG504H (50:50, lactide:glycolide), RG756 (75:25) and mixture of them. They were dissolved in N-methyl-2-pyrrolidone (33% w/w) and mixed with betamethasone acetate (BTMA, 5 and 10% w/w) and ethyl heptanoate
Monographs on fragrance raw materials.
D L Opdyke
Food and cosmetics toxicology, 19(2), 237-254 (1981-04-01)
Supratim Ghosh et al.
Journal of agricultural and food chemistry, 54(5), 1829-1837 (2006-03-02)
Aroma compounds partition between the dispersed and the continuous phases in emulsions, and phase transitions in the lipid droplets profoundly affect the position of the equilibrium. In the present study, the release of ethyl butyrate, ethyl pentanoate, ethyl heptanoate, and
J Enrique Cometto-Muñiz et al.
Experimental brain research, 158(2), 196-206 (2004-04-28)
A series of experiments probed into the degree of chemosensory detection additivity exhibited by mixtures of ethyl propanoate and heptanoate in terms of their trigeminal detectability via nasal pungency (i.e., irritation) and eye irritation. Nasal pungency was tested in subjects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service