Merck
  • Morphine pharmacokinetics and pharmacodynamics in preterm and term neonates: secondary results from the NEOPAIN trial.
Morphine pharmacokinetics and pharmacodynamics in preterm and term neonates: secondary results from the NEOPAIN trial.
British journal of anaesthesia (2008-08-30)
K J S Anand, B J Anderson, N H G Holford, R W Hall, T Young, B Shephard, N S Desai, B A Barton
ABSTRACT
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100 microg kg(-1)), morphine infusions [23-26 weeks postmenstrual age (PMA) 10 microg kg(-1) h(-1); 27-29 weeks 20 microg kg(-1) h(-1); and 30-32 weeks 30 microg kg(-1) h(-1)] were established for a maximum of 14 days. Open-label morphine (20-100 microg kg(-1)) was given for pain or agitation. Morphine assay and neonatal response to ETT suctioning was measured at 20-28 and 70-76 h after starting the drug infusion and at 10-14 h after discontinuation of the study drug. The concentration-effect response was investigated using non-linear mixed effects models. A total of 5119 data points (1598 measured morphine concentrations and 3521 effect measures) were available from 875 neonates for analysis. Clearance was 50% that of the mature value at 54.2 weeks PMA (CLmat(50)) and increased from 2.05 litre h(-1) 70 kg(-1) at 24 weeks PMA to 6.04 litre h(-1) 70 kg(-1) at 32 weeks PMA. The volume of distribution in preterm neonates was 190 litre 70 kg(-1) (CV 51%) and did not change with age. There was no relationship between morphine concentrations (range 0-440 microg litre(-1)) and heart rate changes associated with ETT suctioning or with the Premature Infant Pain Profile. A sigmoid curve describing maturation of morphine clearance is moved to the right in preterm neonates and volume of distribution is increased compared with term neonates. Morphine does not alter the neonatal response to ETT suctioning.
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Pricing and availability is not currently available.
Supelco
Ethyl acetate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Pricing and availability is not currently available.