Skip to Content
Merck
All Photos(1)

Key Documents

W208000

Sigma-Aldrich

Isoamyl octanoate

≥98%, FG

Synonym(s):

Isoamyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
214.34
FEMA Number:
2080
EC Number:
Council of Europe no.:
401
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.120
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.426 (lit.)

bp

260 °C

density

0.861 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coconut; green; fruity; waxy; pineapple; sweet

SMILES string

CCCCCCCC(=O)OCCC(C)C

InChI

1S/C13H26O2/c1-4-5-6-7-8-9-13(14)15-11-10-12(2)3/h12H,4-11H2,1-3H3

InChI key

XKWSWANXMRXDES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoamyl octanoate is an aroma compound found in different kinds of wine.

Biochem/physiol Actions

Taste at 20 ppm

Other Notes

Natural occurrence: Scotch, whiskey, bourbon whiskey, wine, banana, apple and strawberry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds by SPME-GC as age markers of sparkling wines.
Francioli S, et al.
American Journal of Enology and Viticulture, 54(3), 158-162 (2003)
Determination of volatile aroma compounds of Blaufrankisch wines extracted by solid-phase microextraction.
Vas G, et al.
Journal of Chromatographic Science, 36(10), 505-510 (1998)
Quantitative determination of the odorants of young red wines from different grape varieties.
Ferreira V, et al.
Journal of the Science of Food and Agriculture, 80(11), 1659-1667 (2000)
Mosciano, G.
Perfumer & Flavorist, 24, 50-50 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service