Skip to Content
Merck
All Photos(1)

Key Documents

W243205

Sigma-Aldrich

Ethyl decanoate

≥98%, FCC, FG

Synonym(s):

Capric acid ethyl ester, Ethyl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOC2H5
CAS Number:
Molecular Weight:
200.32
FEMA Number:
2432
Beilstein:
1762128
EC Number:
Council of Europe no.:
309
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.059
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

6.9 (vs air)

Assay

≥98%

refractive index

n20/D 1.425 (lit.)

bp

245 °C (lit.)

density

0.865 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; fruity; waxy; sweet

SMILES string

CCCCCCCCCC(=O)OCC

InChI

1S/C12H24O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h3-11H2,1-2H3

InChI key

RGXWDWUGBIJHDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumin Lee et al.
Colloids and surfaces. B, Biointerfaces, 47(1), 78-84 (2006-01-13)
The relationship between the kinetics of the lipase-catalyzed oil hydrolysis and the surface area distribution of oil droplets was investigated using ethyl decanoate and gum Arabic (GA) as a model oil and an emulsifier, respectively. Along an ethyl decanoate concentration
J Hilton et al.
Pharmaceutical research, 11(10), 1396-1400 (1994-10-01)
We studied the effects of three vehicles (propylene glycol, octanol and ethyl decanoate) with differing polarity on the in vitro percutaneous absorption of three chemicals (fluazifop-butyl, dimethyl phthalate and fomesafen sodium salt) with a range of physico-chemical properties. Absorption rate
Magdalena Gallart et al.
Journal of agricultural and food chemistry, 50(24), 7042-7045 (2002-11-14)
The influence of fatty acids (free and bound as ethyl esters) on wine foaming was studied in different white wines and the corresponding sparkling wines. Moreover, from three of these wines the foam formed by CO(2) injection was separated, and
F B Whitfield et al.
The Journal of dairy research, 67(4), 561-569 (2000-01-11)
Analysis by gas chromatography-mass spectrometry of pasteurized milk with a fruity (pineapple like) off odour and a sour, rancid and soapy taste indicated the presence of concentrations at microg/ml levels of ethyl butanoate, ethyl hexanoate, ethyl octanoate, ethyl decanoate, octanoic
Xin-Zheng Huang et al.
Scientific reports, 5, 11867-11867 (2015-07-08)
In response to insect herbivory, plants emit elevated levels of volatile organic compounds for direct and indirect resistance. However, little is known about the molecular and genomic basis of defense response that insect herbivory trigger in cotton plants and how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service