Skip to Content
Merck
All Photos(2)

Key Documents

T8752

Sigma-Aldrich

Tetracosane

99%

Synonym(s):

n-Tetracosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)22CH3
CAS Number:
Molecular Weight:
338.65
Beilstein:
1758462
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

bp

391 °C (lit.)

mp

49-52 °C (lit.)

solubility

chloroform: soluble 10%, clear, colorless

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C24H50/c1-3-5-7-9-11-13-15-17-19-21-23-24-22-20-18-16-14-12-10-8-6-4-2/h3-24H2,1-2H3

InChI key

POOSGDOYLQNASK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetracosane is a linear alkane. Structural characteristics of confined squalane and tetracosane under shear flow conditions have been investigated.[1] Solubility of tetracosane in ethane and ethylene have been investigated.[2]
Viscosity, liquid density, and surface tension data of its binary and ternary mixtures with various n-alkanes (n-heptane, n-decane or n-hexadecane) have been evaluated.[3] Its solubility in various hydrocarbon solvents (heptane, dodecane, isomeric mixtures of decahydronapthalene, m- and p-xylene) has been studied.[4]

Application

Tetracosane is the suitable solvent used in the synthesis of ZnS nanoparticle.[5] It may be employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasun Mukherjee et al.
The journal of physical chemistry. A, 115(16), 4031-4041 (2010-11-26)
This work explores the sensitization of luminescent lanthanide Tb(3+) and Eu(3+) cations by the electronic structure of zinc sulfide (ZnS) semiconductor nanoparticles. Excitation spectra collected while monitoring the lanthanide emission bands reveal that the ZnS nanoparticles act as an antenna
Antía Lestido-Cardama et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-23)
Dialkyl diketene dimers are used as sizing agents in the manufacture of paper and board for food contact applications to increase wetting stability. Unbound residues can hydrolyze and decarboxylate into dialkylketones. These non-intentionally added substances (NIAS) have potential to migrate
How polymer additives reduce the pour point of hydrocarbon solvents containing wax crystals.
Binks BP, et al.
Physical Chemistry Chemical Physics (2015)
Viscosity and liquid density of asymmetric hydrocarbon mixtures.
Queimada AJ, et al.
International Journal of Thermophysics, 24(5), 1221-1239 (2003)
Shear behavior of squalane and tetracosane under extreme confinement. I. Model, simulation method, and interfacial slip.
Gupta SA, et al.
J. Chem. Phys. , 107(23), 10316-10326 (1997)

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service