Skip to Content
Merck
All Photos(1)

Key Documents

8.20547

Sigma-Aldrich

n-Eicosane

for synthesis

Synonym(s):

n-Eicosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18CH3
CAS Number:
Molecular Weight:
282.55
MDL number:
UNSPSC Code:
12352001
EC Index Number:
204-018-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.001 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

204-206 °C/20 hPa

mp

34-37 °C

transition temp

flash point 176 °C

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCCCCC)CCCCCCCCC

InChI

1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3

InChI key

CBFCDTFDPHXCNY-UHFFFAOYSA-N

Application

  • Effect of temperature and graphite particle fillers on thermal conductivity and viscosity of phase change material n-eicosane: This study discusses the enhanced thermal conductivity and viscosity changes of n-eicosane when mixed with graphite particle fillers, important for applications in energy storage and thermal management (Srinivasan et al., 2017).
  • Self-Assembly Synthesis of Microencapsulated n-Eicosane Phase-Change Materials with Crystalline-Phase-Controllable Calcium Carbonate Shell: This paper covers the synthesis of microencapsulated n-eicosane with a calcium carbonate shell, crucial for enhancing thermal energy storage systems in drug delivery mechanisms (Yu et al., 2014).
  • Preparation and thermal properties of n-eicosane/nano-SiO2/expanded graphite composite phase-change material for thermal energy storage: This research explores the preparation and enhanced thermal properties of n-eicosane composites, beneficial for applications in material sciences focusing on energy efficiency (Zhang et al., 2020).
  • An experimental study of enhanced heat sinks for thermal management using n-eicosane as phase change material: This article evaluates the use of n-eicosane in enhancing heat sinks, which is significant for designing more efficient cooling systems in pharmaceutical manufacturing processes (Arshad et al., 2018).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 36 °C
Melting range (upper value): ≤ 39 °C
Identity (GC): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service