T54607
1,3,5-Trichlorobenzene
99%
Synonym(s):
Symmetrical trichlorobenzene, s-Trichlorobenzene
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H3Cl3
CAS Number:
Molecular Weight:
181.45
Beilstein:
1635233
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder or crystals
bp
208 °C (lit.)
mp
56-60 °C (lit.)
SMILES string
Clc1cc(Cl)cc(Cl)c1
InChI
1S/C6H3Cl3/c7-4-1-5(8)3-6(9)2-4/h1-3H
InChI key
XKEFYDZQGKAQCN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1,3,5-Trichlorobenzene can be used to synthesize:
- Bis(3,5-dichloro-4-pyridyl)(2,4,6-trichlorophenyl)methyl radical, a stable, photoluminescent organic radical.
- Solution-processable graphene quantum dots as light absorbers for photovoltaics.
- π-conjugated polydentate ligand, hexakis(2-pyridyl)dipyrazinoquinoxaline (HPDQ) as a highly selective on/off fluorescence sensor for cadmium (II) ions.
- Two-dimensional chlorine-substituted graphdiyne (Cl-GDY) as an electrode material for lithium-ion batteries.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Large, solution-processable graphene quantum dots as light absorbers for photovoltaics.
Yan X, et al.
Nano Letters, 10(5), 1869-1873 (2010)
A luminescent organic radical with two pyridyl groups: high photostability and dual stimuli-responsive properties, with theoretical analyses of photophysical processes.
Kimura S, et al.
Chemical Science, 9(7), 1996-2007 (2018)
Synthesis of Chlorine?Substituted Graphdiyne and Applications for Lithium?Ion Storage.
Wang N, et al.
Angewandte Chemie (International Edition in English), 56(36), 10740-10745 (2017)
A highly selective on/off fluorescence sensor for cadmium (II).
Zhao Q, et al.
Inorganic Chemistry, 50(20), 10041-10046 (2011)
Julian Renpenning et al.
Rapid communications in mass spectrometry : RCM, 31(13), 1095-1102 (2017-04-05)
The conventional high-temperature conversion (HTC) approach towards hydrogen compound-specific isotope analysis (CSIA) of halogen-bearing (F, Cl, Br, I) organics suffers from incomplete H The performance of the modified gas chromatography/chromium-based high-temperature conversion (GC-Cr/HTC) system was monitored and optimized using an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service