Skip to Content
Merck
All Photos(3)

Documents

T2207

Sigma-Aldrich

Terephthalaldehyde

ReagentPlus®, 99%

Synonym(s):

Benzene-1,4-dicarboxaldehyde, Terephthaldicarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CHO)2
CAS Number:
Molecular Weight:
134.13
Beilstein:
385863
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

bp

245-248 °C (lit.)

mp

114-116 °C (lit.)

SMILES string

[H]C(=O)c1ccc(cc1)C([H])=O

InChI

1S/C8H6O2/c9-5-7-1-2-8(6-10)4-3-7/h1-6H

InChI key

KUCOHFSKRZZVRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Terephthalaldehyde can be used as a starting material for:
  • The synthesis of fused bis-benzoquinoline derivatives on reacting with arylamines and cyclic ketones through one pot three component reaction.
  • The photochemical synthesis of 5,10-disubstituted[5]helicenes by reacting with benzyl cyanide via Knoevenagel reaction.
  • The synthesis of a nitronyl nitroxide (NIT) radical with a pyrimidiniumolate unit, which are useful precursors for building molecular magnets.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Facile photochemical synthesis of 5, 10-disubstituted [5] Helicenes by removing molecular orbital degeneracy
Ito N, et al.
Organic Letters, 16(9), 2502-2505 (2014)
Synthesis of dipolar nitronyl nitroxides
Greve S, et al.
Organic Letters, 2(15), 2269-2270 (2000)
Camphorsulfonic acid catalyzed one-pot three-component reaction for the synthesis of fused quinoline and benzoquinoline derivatives
Gattu R, et al.
The Journal of Organic Chemistry, 82(23), 12416-12429 (2017)
Niklas Keller et al.
Nanoscale, 11(48), 23338-23345 (2019-12-04)
Covalent organic frameworks (COFs), consisting of covalently connected organic building units, combine attractive features such as crystallinity, open porosity and widely tunable physical properties. For optoelectronic applications, the incorporation of heteroatoms into a 2D COF has the potential to yield
Chenyu Lin et al.
BMC chemistry, 14(1), 66-66 (2020-12-10)
For analysis of weak π-π complexes proton-nuclear magnetic resonance (proton-NMR) simultaneously provides information of stacking configurations and association constants [Formula: see text] However, an apparent issue for this approach is inconsistent/impossible constant estimation which often leads to unreasonable interpretation for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service