Skip to Content
Merck
All Photos(1)

Documents

753491

Sigma-Aldrich

Benzene-1,3,5-tricarboxaldehyde

97%

Synonym(s):

1,3,5-Triformylbenzene, Trimesaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

156-161 °C

storage temp.

2-8°C

SMILES string

O=Cc1cc(C=O)cc(C=O)c1

InChI

1S/C9H6O3/c10-4-7-1-8(5-11)3-9(2-7)6-12/h1-6H

InChI key

AEKQNAANFVOBCU-UHFFFAOYSA-N

Application

Benzene-1,3,5-tricarboxaldehyde is extensively used in the synthesis of a wide range of porous organic cages and covalent organic frameworks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingming Hua et al.
ACS nano, 14(5), 5517-5528 (2020-05-07)
Although self-assembly across multiple length scales has been well recognized and intensively investigated in natural biological system, the design of artificial heterostructures enabled by integrative self-assembly is still in its infancy. Here we report a strategy toward the growth of
Lilia Clima et al.
Polymers, 11(8) (2019-08-09)
Polyethylene glycol (PEG) functionalization of non-viral vectors represents a powerful tool through the formation of an overall surface charge shielding ability, which is fundamental for efficient nucleic acid delivery systems. The degree of non-viral vector PEGylation and the molecular weight
Two fully conjugated covalent organic frameworks as anode materials for lithium ion batteries.
Bai L, et al.
Journal of Material Chemistry A, 4(37), 14106-14110 (2016)
Helical Rod-like Phenylene Cages via Ruthenium Catalyzed Diol-Diene Benzannulation: A Cord of Three Strands.
Sato H, et al.
Journal of the American Chemical Society, 140(7), 2455-2459 (2018)
Shan Jiang et al.
Nanoscale, 11(31), 14929-14936 (2019-07-31)
Porous Organic Cages (POCs) are an emerging class of self-assembling, porous materials with novel properties. They offer a key advantage over other porous materials in permitting facile solution processing and re-assembly. The combination of POCs with metal nanoparticles (NPs) unlocks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service