Skip to Content
Merck
All Photos(1)

Key Documents

T101

Sigma-Aldrich

Gaboxadol hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

4,5,6,7-Tetrahydroisoxazolo[5,4-c]pyridin-3-ol hydrochloride, THIP hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2 · HCl
CAS Number:
Molecular Weight:
176.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

ethanol: 0.91 mg/mL
H2O: 20 mg/mL

SMILES string

Cl[H].Oc1noc2CNCCc12

InChI

1S/C6H8N2O2.ClH/c9-6-4-1-2-7-3-5(4)10-8-6;/h7H,1-3H2,(H,8,9);1H

InChI key

ZDZDSZQYRBZPNN-UHFFFAOYSA-N

General description

Gaboxadol, also referred to as 4,5,6,7-tetrahydroisoxazolo(5,4-c)pyridin-3-ol (THIP), is a GABA site agonist. It has great affinity to the δ subunit of extrasynaptic high-affinity GABAA receptors. Gaboxadol was first developed for the treatment of schizophrenia, epilepsy, primary insomnia and Huntington′s disease. It can easily cross the blood-brain barrier and is rapidly absorbed by the body.

Application

Gaboxadol hydrochloride has been used in cell migration and invasion assay to assess liver cancer cell migration.

Biochem/physiol Actions

GABAA receptor agonist.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Krogsgaard-Larsen et al.
Journal of medicinal chemistry, 26(6), 895-900 (1983-06-01)
The thio analogues of the GABA (gamma-aminobutyric acid) agonist THIP (4,5,6,7-tetrahydroisoxazolo[5,4-c]pyridin-3-ol), the GABA uptake inhibitor THPO (4,5,6,7-tetrahydroisoxazolo[4,5-c]pyridin-3-ol), and the glycine antagonist THAZ (5,6,7,8-tetrahydro-4H-isoxazolo[4,5-d]azepin-3-ol) have been synthesized and tested biologically on single neurons in the cat spinal cord and in vitro
GABA(A) agonists and partial agonists: THIP (Gaboxadol) as a non-opioid analgesic and a novel type of hypnotic.
Krogsgaard-Larsen P, et al.
Biochemical Pharmacology, 68(8), 1573-1580 (2004)
GABA and Sleep: Molecular, Functional and Clinical Aspects (2010)
M Lancel
Sleep, 22(1), 33-42 (1999-02-16)
This paper reviews the sleep effects of systemically administered agonistic modulators of GABAA receptors, including barbiturates, benzodiazepines, zolpidem, zopiclone and neuroactive steroids, and the selective GABAA agonists muscimol and THIP. To assess the involvement of GABAA receptors in the physiologic
Vanessa M Hill et al.
PLoS biology, 16(7), e2005206-e2005206 (2018-07-13)
Although sleep appears to be broadly conserved in animals, the physiological functions of sleep remain unclear. In this study, we sought to identify a physiological defect common to a diverse group of short-sleeping Drosophila mutants, which might provide insight into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service