SYNC121120
Synthware™ inner coil condenser
joint: ST/NS 14/20, hose size 8 mm
Synonym(s):
Synthware C121120, Condenser coil, Coil reflux condenser
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C121120
hose size
8 mm
Related Categories
General description
Synthware™ coiled condenser features a 120 mm jacketed length for superior cooling efficiency. With standardized 14/20 upper and lower joints, it easily fits various laboratory glassware. The inner coil circulates coolant, maximizing heat exchange. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)