P55805
Purine
98%
Synonym(s):
7H-Imidazo(4,5-d)pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H4N4
CAS Number:
Molecular Weight:
120.11
Beilstein:
3200
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
214-217 °C (lit.)
SMILES string
c1ncc2nc[nH]c2n1
InChI
1S/C5H4N4/c1-4-5(8-2-6-1)9-3-7-4/h1-3H,(H,6,7,8,9)
InChI key
KDCGOANMDULRCW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Purine is a heterocyclic organic compound constituting a pyrimidine ring fused to an imidazole ring.
Application
Purine can undergo direct C-H functionalization in the presence of palladium catalyst to form various biologically important products.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Benshang Li et al.
Nature medicine, 21(6), 563-571 (2015-05-12)
Relapse is the leading cause of mortality in children with acute lymphoblastic leukemia (ALL). Among chemotherapeutics, thiopurines are key drugs in ALL combination therapy. Using whole-exome sequencing, we identified relapse-specific mutations in the phosphoribosyl pyrophosphate synthetase 1 gene (PRPS1), which
New developments in direct functionalization of C-H and N-H bonds of purine bases via metal catalyzed cross-coupling reactions.
Abdoli M, et al.
Royal Society of Chemistry Advances, 55(5), 44371-44389 (2015)
Hiroshi Ashihara et al.
Phytochemistry, 69(4), 841-856 (2007-12-11)
Details of the recently elucidated biosynthetic pathways of caffeine and related purine alkaloids are reviewed. The main caffeine biosynthetic pathway is a sequence consisting of xanthosine-->7-methylxanthosine-->7-methylxanthine-->theobromine-->caffeine. Genes encoding N-methyltransferases involved in three of these four reactions have been isolated and
Silvia Meneghesso et al.
ChemMedChem, 8(3), 415-425 (2013-02-07)
2'-Fluoro-2'-deoxyguanosine has been reported to have potent anti-influenza virus activity in vitro and in vivo. Herein we describe the synthesis and biological evaluation of 6-modified 2'-fluoro-2'-deoxyguanosine analogues and their corresponding phosphoramidate ProTides as potential anti-influenza virus agents. Whereas the parent
Elijah Oyoo-Okoth et al.
Aquatic toxicology (Amsterdam, Netherlands), 124-125, 34-40 (2012-08-14)
Infestation of fish by endoparasites may potentially influence metal uptake and elimination by the host. We quantified the metal uptake rate constant (k(u)) and efflux rate constants (k(e)) of radiolabeled Cd and Co in the cyprinid fish Rastrineobola argentea experimentally
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service