Skip to Content
Merck
All Photos(1)

Key Documents

C122106

Sigma-Aldrich

Cytidine

99%

Synonym(s):

Cytosine β-D-riboside, Cytosine-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Beilstein:
89173
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

optical activity

[α]20/D +33°, c = 2 in H2O

mp

210-220 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7-,8-/m1/s1

InChI key

UHDGCWIWMRVCDJ-XVFCMESISA-N

Gene Information

mouse ... Uck1(22245)

Looking for similar products? Visit Product Comparison Guide

General description

Cytidine also known as cytosine β-D-riboside, is a nucleoside, commonly used in organic synthesis.

Application

Cytidine can be used as a building block to synthesize 5‘-hydroxy-5‘-phosphonate derivatives of cytidine. Additionally, it is used to regulate the phosphatidate phosphatase activity by nucleotides. [1] [2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of the 5 `-hydroxy-5 `-phosphonate derivatives of cytidine and cytosine arabinoside
X Chen
The Journal of Organic Chemistry, 67, 9331-9339 (2002)
Regulation of phosphatidate phosphatase activity from the yeast Saccharomyces cerevisiae by nucleotides.
The Journal of Biological Chemistry, 269, 29495-29501 (1994)
Koji Hayakawa et al.
The Journal of biological chemistry, 288(24), 17099-17110 (2013-04-30)
The orexin system plays a central role in the integration of sleep/wake and feeding behaviors in a broad spectrum of neural-metabolic physiology. Orexin-A and orexin-B are produced by the cleavage of prepro-orexin, which is encoded on the Hcrt gene. To
G P Gerrits et al.
Clinical chemistry, 34(7), 1439-1442 (1988-07-01)
Disturbances in the metabolism of purines and pyrimidines in neurologically affected patients can be reflected by aberrant concentrations of nucleosides and nucleobases in cerebrospinal fluid (CSF). However, normal values, especially for children at different ages, are lacking. We collected 1000
Takayuki Ohira et al.
The Journal of biological chemistry, 288(11), 7645-7652 (2013-01-31)
Non-universal genetic codes are frequently found in animal mitochondrial decoding systems. In squid mitochondria, four codons deviate from the universal genetic code, namely AUA, UGA, and AGA/AGG (AGR) for Met, Trp, and Ser, respectively. To understand the molecular basis for

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service