P39001
Phthalazone
99%
Synonym(s):
Benzo[d]pyridazin-1(2H)one
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H6N2O
CAS Number:
Molecular Weight:
146.15
Beilstein:
3654
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
mp
183-185 °C (lit.)
SMILES string
O=C1NN=Cc2ccccc12
InChI
1S/C8H6N2O/c11-8-7-4-2-1-3-6(7)5-9-10-8/h1-5H,(H,10,11)
InChI key
IJAPPYDYQCXOEF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Craig Zetterberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(8), 1286-1295 (2016-06-15)
(R)-2-((2-(1H-pyrrolo[2,3-b]pyridin-3-yl)pyrimidin-4-yl)amino)-2-methyl-N-(2,2,2-trifluoroethyl)butanamide (VX-509, decernotinib) is an oral Janus kinase 3 inhibitor that has been studied in patients with rheumatoid arthritis. Patients with rheumatoid arthritis often receive multiple medications, such as statins and steroids, to manage the signs and symptoms of comorbidities
Raghava Choudary Palacharla et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(2), 169-176 (2018-02-01)
The objective is to evaluate methoxsalen as an in vitro phenotyping tool in comparison to ABT as a nonspecific inactivator of P450 mediated metabolism. The reversible inhibition of methoxsalen and ABT against the P450, FMO, AO, MAO-A and -B, enzymes
Yasuhiro Uno et al.
Xenobiotica; the fate of foreign compounds in biological systems, 51(4), 494-499 (2021-01-13)
The cynomolgus macaque is a non-human primate species widely used in drug metabolism studies. Despite the importance of genetic polymorphisms in cytosolic aldehyde oxidase (AOX) 1 in humans, genetic variants have not been investigated in cynomolgus or rhesus macaques.Genetic variants
Takayuki Amano et al.
Biochemical pharmacology, 151, 69-78 (2018-03-10)
Dantrolene is used for malignant hyperthermia during anesthesia, and it sometimes causes severe liver injury as a side effect. Dantrolene is metabolized to acetylaminodantrolene, which is formed via the reduction of dantrolene to aminodantrolene and subsequent acetylation. Formation of hydroxylamine
Shengyun Huang et al.
Journal of separation science, 43(17), 3565-3572 (2020-07-07)
Capillary electrophoresis is a modern separation technique characterized by many benefits, which qualify it also for enzyme assays and the study of enzyme kinetics during drug development. Homogeneous or heterogeneous approaches can be followed for the enzymatic incubation. In this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service