Skip to Content
Merck
All Photos(1)

Documents

T32409

Sigma-Aldrich

2-Thiophenecarboxaldehyde

98%

Synonym(s):

Thenaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4OS
CAS Number:
Molecular Weight:
112.15
Beilstein:
105819
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.591 (lit.)

bp

198 °C (lit.)
75-77 °C/11 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccs1

InChI

1S/C5H4OS/c6-4-5-2-1-3-7-5/h1-4H

InChI key

CNUDBTRUORMMPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anupama Kumari et al.
Chemistry, an Asian journal, 14(13), 2278-2290 (2019-05-08)
A tandem IBX-promoted oxidation of primary alcohol to aldehyde and opening of intermediate β,γ-diolcarbonate aldehyde to (E)-γ-hydroxy-α,β-enal has been developed. Remarkably, the carbonate opening delivered exclusively (E)-olefin and no over-oxidation of γ-hydroxy was observed. The method developed has been extended
Talat Baran
International journal of biological macromolecules, 127, 232-239 (2019-01-15)
One of the most important problems in catalytic systems is metal leaching due to weak interactions between the supporting material and the metal ions. It adversely affects the reusability and catalytic performance of catalysts. In this study, a novel support
Diana Rogacz et al.
Materials (Basel, Switzerland), 12(12) (2019-06-27)
The aim of this work was to evaluate the impact of the thiophene-derived aminophosphonates 1-6 on seedling emergence and growth of monocotyledonous oat (Avena sativa) and dicotyledonous radish (Raphanus sativus L.), and phytotoxicity against three persistent and resistant weeds (Galinsoga
Justine E Giffen-Lemieux et al.
Journal of medical entomology, 57(4), 994-1005 (2020-02-14)
The attraction of necrophagous insects, particularly blow flies, to corpses and carrion is of ecological, economic, and agricultural importance, although the mechanisms by which it occurs are not well understood. Much of the published research on blow fly attractants has
Dahlia Salman et al.
Journal of breath research, 15(1), 016004-016004 (2020-10-27)
Radiation dose is important in radiotherapy. Too little, and the treatment is not effective, too much causes radiation toxicity. A biochemical measurement of the effect of radiotherapy would be useful in personalisation of this treatment. This study evaluated changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service