Skip to Content
Merck
All Photos(1)

Key Documents

P30004

Sigma-Aldrich

1-Phenylpiperazine

≥97.0%

Synonym(s):

N-Phenyldiethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Beilstein:
132157
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.588 (lit.)

bp

286 °C (lit.)

density

1.062 g/mL at 25 °C (lit.)

SMILES string

C1CN(CCN1)c2ccccc2

InChI

1S/C10H14N2/c1-2-4-10(5-3-1)12-8-6-11-7-9-12/h1-5,11H,6-9H2

InChI key

YZTJYBJCZXZGCT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

284.0 °F

Flash Point(C)

140 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jadwiga Handzlik et al.
Bioorganic & medicinal chemistry, 16(11), 5982-5998 (2008-05-21)
In the search for new antiarrhythmic agents, some active 2-methoxyphenylpiperazine derivatives of phenytoin were obtained as a chemical modification of compound AZ-99 (3-ethyl-1-[2-hydroxy-3-(4-phenylpiperazin-1-yl)-propyl]-2,4-dioxo-5,5-diphenylimidazolidine). These compounds possessed structural properties similar to those of alpha(1)-adrenoceptor antagonists. In the present study, the affinities
Gürkan Keşan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 144-155 (2012-01-10)
New metal halide complexes in the form of M(pp)(2)Cl(2) (where pp=1-phenylpiperazine and M=Pd or Hg) have been prepared for the first time and their FT-IR and FT-Raman spectra are reported in the region of 4000-10 cm(-1) and 4000-50 cm(-1), respectively.
Tiantian Mou et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(11), 2013-2018 (2009-08-15)
This study reports the synthesis and characterization of N-(3-(4-(2-methoxyphenyl)piperazin-1-yl)propyl-4-[(18)F]fluorobenzamide ([(18)F]MPP3F). The total reaction time for [(18)F]MPP3F, including final high-performance liquid chromatography purification, was about 3h. Typical decay-corrected radiochemical yield was 18.4+/-3.1%. The radiochemical purity was >98%. Biodistribution in mice showed
Sabine Linz et al.
Bioorganic & medicinal chemistry, 17(13), 4448-4458 (2009-06-02)
A series of phenylpiperazine-methyl-substituted 1H-pyrrolo[2,3-c]pyridine, imidazo[1,2-c]-, pyrrolo[2,3-d]- and pyrrolo[3,2-d]pyrimidines were prepared as selective dopamine D4-ligands. The pyrrolo[2,3-d]pyrimidine derivatives 12d (K(i)=1,9 nM) and 34 d (K(i)=2,4 nM) as well as the pyrrolo[3,2-d]pyrimidine Mannich base 49f (K(i)=2,8 nM) showed high dopamine D4
Katharina Ehrlich et al.
Journal of medicinal chemistry, 52(15), 4923-4935 (2009-07-18)
Assembling phenylpiperazines with 7a-azaindole via different spacer elements, we developed subtype selective dopamine receptor ligands of types 1a,c, 2a, and 3a preferentially interacting with D4, D2, and D3, respectively. To complete this set, the methylthio analogues 2b and 3b exceeding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service