Skip to Content
Merck
All Photos(3)

Key Documents

P0256

Sigma-Aldrich

Pyrithiamine hydrobromide

~95%, crystalline

Synonym(s):

1-([4-Amino-2-methyl]-5-pyrimidylmethyl)-2-methyl-3-(β-hydroxyethyl)pyridinium bromide, Neopyrithiamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O · 2HBr
CAS Number:
Molecular Weight:
420.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

~95%

Quality Level

form

crystalline

color

off-white to yellow

storage temp.

2-8°C

SMILES string

Br.[Br-].Cc1ncc(C[n+]2cccc(CCO)c2C)c(N)n1

InChI

1S/C14H19N4O.2BrH/c1-10-12(5-7-19)4-3-6-18(10)9-13-8-16-11(2)17-14(13)15;;/h3-4,6,8,19H,5,7,9H2,1-2H3,(H2,15,16,17);2*1H/q+1;;/p-1

InChI key

DMVBWVASDWGFNH-UHFFFAOYSA-M

General description

Pyrithiamine hydrobromide is a heterocyclic building block that is used as a valuable tool in organic synthesis and medicinal chemistry due to its ability to mimic thiamine deficiency and its potential for the development of new compounds with useful pharmacological properties.

Application

Pyrithiamine hydrobromide can be used as a precursor for the synthesis pyrithiamine pyrophosphate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica J Roland et al.
Neurobiology of learning and memory, 87(1), 123-132 (2006-09-19)
A rodent model of diencephalic amnesia, pyrithiamine-induced thiamine deficiency (PTD), was used to investigate the dynamic role of hippocampal and striatal acetylcholine (ACh) efflux across acquisition of a nonmatching-to-position (NMTP) T-maze task. Changes in ACh efflux were measured in rats
Thiamine pyrophosphate riboswitches are targets for the antimicrobial compound pyrithiamine
Sudarsan, et al.
Chemistry & Biology, 12, 1325-1335 (2005)
Steven Anzalone et al.
The European journal of neuroscience, 32(5), 847-858 (2010-08-24)
Although the key neuropathology associated with diencephalic amnesia is lesions to the thalamus and/or mammillary bodies, functional deactivation of the hippocampus and associated cortical regions also appear to contribute to the memory dysfunction. For example, there is loss of forebrain
Xiaohua He et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 32(10), 2207-2216 (2007-02-15)
The relative roles of alcohol and thiamine deficiency in causing brain damage remain controversial in alcoholics without the Wernicke-Korsakoff syndrome. Experimental control over alcohol consumption and diet are impossible in humans but can be accomplished in animal models. This experiment
Stéphane Thore et al.
Science (New York, N.Y.), 312(5777), 1208-1211 (2006-05-06)
Riboswitches are untranslated regions of messenger RNA, which adopt alternate structures depending on the binding of specific metabolites. Such conformational switching regulates the expression of proteins involved in the biosynthesis of riboswitch substrates. Here, we present the 2.9 angstrom-resolution crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service