Skip to Content
Merck
All Photos(1)

Documents

M46705

Sigma-Aldrich

N-Methylformamide

99%

Synonym(s):

N-Formylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHCH3
CAS Number:
Molecular Weight:
59.07
Beilstein:
1098352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Assay

99%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

198-199 °C (lit.)

mp

−4 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)NC

InChI

1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)

InChI key

ATHHXGZTWNVVOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walther Caminati et al.
Physical chemistry chemical physics : PCCP, 12(35), 10230-10234 (2010-07-27)
We investigated the Fourier transform microwave spectra of the hydrated forms of N-methylformamide (NMF) in a supersonic expansion and assigned the rotational spectra of two mono-hydrated species. The conformation of each molecular complex was reliably determined on the basis of
Edward M Kosower et al.
The journal of physical chemistry. B, 113(16), 5622-5632 (2009-03-31)
N-methylformamide (NMF), the simplest model for peptides, exhibits hyperplectic (both simple and complex) behavior as revealed by thin film infrared spectroscopy on planar AgX [AgCl:AgBr] fiber. IR spectra (0.1 s scans) of 10 microg NMF/dichloromethane(DCM) under N(2) flow first show
Jeppe S Spicker et al.
Toxicological sciences : an official journal of the Society of Toxicology, 102(2), 444-454 (2008-01-08)
A large number of databases are currently being implemented within toxicology aiming to integrate diverse biological data, such as clinical chemistry, expression, and other types of data. However, for these endeavors to be successful, tools for integration, visualization, and interpretation
Jolanta Swiergiel et al.
The journal of physical chemistry. B, 113(43), 14225-14228 (2009-10-07)
Impedance spectroscopy was used for studies of the frequency dependence of the electric conductivity of two highly conducting liquid amides: N-methylformamide (NMF), strongly self-associated via the hydrogen bonds C=O...N-H, and N,N-dimethylformamide (DMF), which is not able to self-organize by hydrogen
Shih-Min Wang et al.
The Science of the total environment, 388(1-3), 398-404 (2007-09-15)
N,N-dimethylformamide (DMF) could be readily absorbed via skin and inhalation routes. It is difficult, however, to separate the internal dose contribution from skin vapor and inhalation exposure. This study attempts to quantitatively determine the separate skin vapor and inhalation exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service