M2101
Mandelic acid
99%
Synonym(s):
(RS)-Mandelic acid, (±)-Mandelic Acid, DL-Mandelic acid, Racemic mandelic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
mp
119-121 °C (lit.)
SMILES string
OC(C(O)=O)c1ccccc1
InChI
1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)
InChI key
IWYDHOAUDWTVEP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Racemic mandelic acid in the form of mandelates is generally used as a pharmaceutical component due to its analgesic, antirheumatic, and spasmolytic effects. It undergoes resolution to form (+)- and (-)-enantiomers that are widely used as chiral resolving agents in enantioseparation of various other racemates via salt formation.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Enantiomeric mandelic acid system melting point phase diagram and solubility in water.
Lorenz H, et al.
Journal of Chemical and Engineering Data, 47(5), 1280-1284 (2002)
P Vodicka et al.
Carcinogenesis, 16(7), 1473-1481 (1995-07-01)
Occupational exposure to styrene was studied in nine workers of a hand lamination plant in Bohemia. Personal dosimeters were used to monitor the styrene workplace exposure, and the levels of styrene in blood and mandelic acid in urine were measured.
Ludmila Krivánková et al.
Electrophoresis, 24(3), 505-517 (2003-02-06)
Effects originating from the variability of the sample matrix can be efficiently eliminated when the separation conditions are selected so that compounds of like charge with high concentration referred to as macrocomponents are embodied into the system of transient isotachophoresis.
David A Ostrov et al.
Antimicrobial agents and chemotherapy, 51(10), 3688-3698 (2007-08-08)
The bacterial type II topoisomerases DNA gyrase and topoisomerase IV are validated targets for clinically useful quinolone antimicrobial drugs. A significant limitation to widely utilized quinolone inhibitors is the emergence of drug-resistant bacteria due to an altered DNA gyrase. To
G L Corona et al.
European archives of psychiatry and neurological sciences, 239(2), 79-86 (1989-01-01)
Memory performance, central monoaminergic function and sympathetic nerve activity were studied in patients with dementia of the Alzheimer type (DAT) or with multi-infarct dementia before and after 4 weeks with single or combined drug therapy (choline-piracetam). Analysis of the levels
Chromatograms
suitable for GCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service