Skip to Content
Merck
All Photos(1)

Key Documents

328022

Sigma-Aldrich

4-Methylhippuric acid

98%

Synonym(s):

N-(4-Methylbenzoyl)glycine, p-Toluric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONHCH2CO2H
CAS Number:
Molecular Weight:
193.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

163-165 °C (lit.)

functional group

amide
carboxylic acid

SMILES string

Cc1ccc(cc1)C(=O)NCC(O)=O

InChI

1S/C10H11NO3/c1-7-2-4-8(5-3-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)

InChI key

NRSCPTLHWVWLLH-UHFFFAOYSA-N

General description

4-Methylhippuric acid is the major metabolite of xylene and has been determined in urine by gas chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P T Mayer et al.
AAPS pharmSci, 2(2), E14-E14 (2001-12-14)
To explore the independence of functional group contributions to permeability of nonelectrolytes across egg lecithin bilayers, the transport rates were measured of a series of alpha-substituted p-methylhippuric acids (-H, -Cl, -OCH3, -CN, -OH, -COOH, and -CONH2) across egg lecithin lipid
G Bieniek et al.
British journal of industrial medicine, 38(3), 304-306 (1981-08-01)
The separation of hippuric and m-methylhippuric acid as toluene and m-xylene metabolites present in urine of people exposed simultaneously to toluene and xylene is described. Chloroform was used for hippuric and m-methylhippuric acid extraction. Satisfactory separation of these metabolites was
Peter T Mayer et al.
Journal of pharmaceutical sciences, 91(3), 640-646 (2002-03-29)
The barrier domain solubility-diffusion theory of lipid bilayer permeability relates the permeability coefficient (P(m)) to the solute's partition coefficient (PC(barrier/w)) and diffusion coefficient (D(barrier)) in the ordered chain region of the bilayer that serves as the barrier region for polar
Measurement of exposure to xylenes by separate determination of m- and p-methylhippuric acids in urine.
M Morin et al.
Journal of chromatography, 210(2), 346-349 (1981-06-05)
Simultaneous determination of hippuric acid and o-, m- and p-methylhippuric acids in urine by high-performance liquid chromatography.
T Sakai et al.
Journal of chromatography, 276(1), 182-188 (1983-08-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service