M14951
5-Methoxyindole-2-carboxylic acid
97%
Synonym(s):
NSC 30927
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9NO3
CAS Number:
Molecular Weight:
191.18
Beilstein:
157478
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
199-201 °C (lit.)
SMILES string
COc1ccc2[nH]c(cc2c1)C(O)=O
InChI
1S/C10H9NO3/c1-14-7-2-3-8-6(4-7)5-9(11-8)10(12)13/h2-5,11H,1H3,(H,12,13)
InChI key
YEBJVSLNUMZXRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Reactant for preparation of:
- Anticancer agents
- A fluorescent small molecule probe for in vivo lipid imaging
- Indoleamine 2,3-dioxygenase (IDO) inhibitors
- Selective Dopamine D3 receptor ligands
- 5-HT4 receptor ligands
- Inhibitors of Mycobacterium tuberculosis pantothenate synthetase
- Hypoxia selective cytotoxins
- Potent antitumor bifunctional DNA alkylating agents
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nathalie Sumien et al.
Behavioural brain research, 378, 112278-112278 (2019-10-21)
Stroke leads to devastating outcomes including impairments of sensorimotor and cognitive function that may be long lasting. New intervention strategies are needed to overcome the long-lasting effects of ischemic injury. Previous studies determined that treatment with 5-methoxyindole-2-carboxylic acid (MICA) conferred
Jinzi Wu et al.
Biochemical and biophysical research communications, 497(1), 444-450 (2018-02-16)
We previously reported that 5-methoxyindole-2-carboxylic acid (MICA) could induce preconditioning effect in the ischemic brain of rat. In the present study, we addressed the question of whether MICA could also trigger a postconditioning effect in ischemic stroke. To this end
Luna Yamamori et al.
Zoological science, 34(3), 173-178 (2017-06-08)
Scyphozoa, Cubozoa and Hydrozoa are classes in the phylum Cnidaria that undergo metagenesis involving a dramatic morphological transition. In Scyphozoa and Cubozoa, when exposed to species- or strain-specific transition-inducing stimuli, asexually reproducing benthic polyps transform into sexually reproducing planktonic medusae.
Jinzi Wu et al.
Free radical biology & medicine, 113, 244-254 (2017-10-12)
The objective of this study was to investigate a possible role of mitochondrial dihydrolipoamide dehydrogenase (DLDH) as a chemical preconditioning target for neuroprotection against ischemic injury. We used 5-methoxyindole-2-carboxylic acid (MICA), a reportedly reversible DLDH inhibitor, as the preconditioning agent
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service