Skip to Content
Merck
All Photos(4)

Key Documents

I5109

Sigma-Aldrich

Indole-2-carboxylic acid

98%

Synonym(s):

2-Carboxyindole, 2-Indolylformic acid, NSC 16598

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
124132
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

202-206 °C (lit.)

SMILES string

OC(=O)c1cc2ccccc2[nH]1

InChI

1S/C9H7NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h1-5,10H,(H,11,12)

InChI key

HCUARRIEZVDMPT-UHFFFAOYSA-N

Gene Information

human ... SRD5A1(6715)
rat ... Grin2a(24409)

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for total synthesis of (±)-dibromophakellin and analogs
  • Reactant for synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine
  • Reactant for stereoselective preparation of renieramycin G analogs
  • Reactant for preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization
  • Reactant for Pd-catalyzed cyclization
  • Reactant for preparation of N,N′-(pentane)diylbis[indolecarboxamide] and N,N′-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Kuehm-Caubere et al.
Journal of medicinal chemistry, 40(8), 1201-1210 (1997-04-11)
Series of indole-2-carboxamide and cycloalkeno[1,2-b]indole derivatives were synthesized and evaluated in order to determine the necessary structural requirements for a high inhibition of human LDL copper-induced peroxidation. Various modulations were systematically performed on the indole and cycloalkeno[1,2-b]indole nuclei as well
Stephen P Nighswander-Rempel et al.
Photochemistry and photobiology, 84(3), 613-619 (2008-01-23)
We have synthesized a compound ideally suited to the study of structure-function relationships in eumelanin synthesis. N-methyl-5-hydroxy-6-methoxy-indole (MHMI) has key functional groups strategically placed on the indole framework to hinder binding in the 2, 5, 6 and 7 positions. Thus
R Di Fabio et al.
Journal of medicinal chemistry, 40(6), 841-850 (1997-03-14)
A series of indole-2-carboxylates bearing suitable chains at the C-3 position of the indole nucleus was synthesized and evaluated in terms of in vitro affinity using [3H]glycine binding assay and in vivo potency by inhibition of convulsions induced by N-methyl-D-aspartate
Hideyuki Shiozawa et al.
Journal of the American Chemical Society, 124(15), 3914-3919 (2002-04-11)
Glycopeptide antibiotics of the vancomycin group bind to bacterial cell wall analogue precursors, and typically also form dimers. We have studied the interplay between these two sets of noncovalent bonds formed at separate interfaces. Indole-2-carboxylic acid (L) forms a set
R Di Fabio et al.
Journal of medicinal chemistry, 42(18), 3486-3493 (1999-09-10)
A series of analogues of the indole-2-carboxylate GV150526, currently in clinical trials as a potential neuroprotective agent for the control of the cerebral damage after stroke onset, was designed based on previous studies dealing with the electronic features of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service