Skip to Content
Merck
All Photos(3)

Key Documents

I5133

Sigma-Aldrich

Imidazolidinyl urea

Synonym(s):

N,N′′-Methylenebis[N′-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N8O8
CAS Number:
Molecular Weight:
388.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

240.5 °C

solubility

water: soluble 1 g/L at 20 °C

density

1.24 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O]
OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O

InChI

[1S/C11H16N8O8/c20-2-18-4(6(22)16-10(18)26)14-8(24)12-1-13-9(25)15-5-7(23)17-11(27)19(5)3-21/h4-5,20-21H,1-3H2,(H2,12,14,24)(H2,13,15,25)(H,16,22,26)(H,17,23,27)]
1S/C11H16N8O8/c20-2-18-4(6(22)16-10(18)26)14-8(24)12-1-13-9(25)15-5-7(23)17-11(27)19(5)3-21/h4-5,20-21H,1-3H2,(H2,12,14,24)(H2,13,15,25)(H,16,22,26)(H,17,23,27)

InChI key

[ZCTXEAQXZGPWFG-UHFFFAOYSA-N]
ZCTXEAQXZGPWFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Imidazolidinyl urea can be used as a hydrogen-bonding reinforced factor to prepare:
  • Polymeric supramolecular hydrogels by using polyethyleneglycol (PEG) and diisocyanates.
  • Multi-functional polyacrylamide hydrogels.
Used in:
Pharmacological studies investigating nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis
Therapeutic studies involving nanostructured vegetable-based carriers for topical delivery of active molecules
Topical formulas for use as antioxidants

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marléne Isaksson et al.
Contact dermatitis, 54(1), 29-34 (2006-01-24)
A topical corticosteroid preparation on the Swedish market, Flutivate cream, contains a fairly high concentration of formaldehyde (FA). In this study, we have investigated the clinical relevance of contact allergy to FA when treating an allergic eczema with Flutivate cream
D Matosiuk et al.
European journal of medicinal chemistry, 36(10), 783-797 (2001-12-12)
The synthesis and physicochemical properties of new carbonyl derivatives of 1-aryl-2-iminoimidazolidine are presented. Isomeric 1-(1-arylimidazolidine-2-ylidene)-3-arylureas (series A) and 1-aryl-2-imine-3-arylaminocarbonylimidazolidines (series B) were obtained after the condensation reaction of 1-aryl-2-iminoimidazolidines and arylisocyanates. 1-Aryl-2-iminoimidazolidines were synthesised in a two-step reaction from the
Onjeon Ryu et al.
Biomolecules & therapeutics, 26(6), 608-615 (2018-02-13)
Benzalkonium chloride, diazolidinyl urea, and imidazolidinyl urea are commonly used preservatives in cosmetics. Recent reports suggested that these compounds may have cellular and systemic toxicity in high concentration. In addition, diazolidinyl urea and imidazolidinyl urea are known formaldehyde (FA) releasers
Robert L Rietschel et al.
Dermatitis : contact, atopic, occupational, drug, 18(3), 155-162 (2007-08-30)
To determine whether petrolatum or aqueous vehicles are more sensitive for detecting allergy to imidazolidinylurea (IU), diazolidinylurea (DU), and dimethylol dimethyl hydantoin (DM). The relationship of these allergens to formaldehyde sensitivity was also explored. Retrospective analysis of patients patch-tested by
T Agner et al.
Contact dermatitis, 45(1), 21-25 (2001-06-26)
The preservatives imidazolidinyl urea (IMID, Germall 115) and diazolidinyl urea (DU, Germall II) are commonly used in cosmetic products and are well-known sensitizers. The aim of the present study was to establish the optimal patch test concentration in hydrophilic dried-in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service