Skip to Content
Merck
All Photos(1)

Key Documents

307270

Sigma-Aldrich

Benzofuran-2-carboxylic acid

99%

Synonym(s):

Coumarilic acid, Coumarone-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Beilstein:
124204
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

mp

193-196 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cc2ccccc2o1

InChI

1S/C9H6O3/c10-9(11)8-5-6-3-1-2-4-7(6)12-8/h1-5H,(H,10,11)

InChI key

OFFSPAZVIVZPHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzofuran-2-carboxylic acid was used in the synthesis of the oxymethyl-modified coumarinic acid-based cyclic DADLE (H-Tyr-D-Ala-Gly-Phe-D-Leu-OH) prodrug[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Ouyang et al.
The journal of peptide research : official journal of the American Peptide Society, 59(4), 183-195 (2002-04-26)
The coumarinic acid-based cyclic DADLE (H-Tyr-D-Ala-Gly-Phe-D-Leu-OH) prodrug 1a exhibited more favorable physicochemical properties than did DADLE for permeation across the intestinal mucosa. However, prodrug 1a, whose bioconversion to DADLE was slow, was subject to extensive biliary clearance when administered to
Yibin Xiang et al.
Bioorganic & medicinal chemistry letters, 21(10), 3050-3056 (2011-04-22)
Novel benzofuran-2-carboxylic acids, exemplified by 29, 38 and 39, have been discovered as potent Pim-1 inhibitors using fragment based screening followed by X-ray structure guided medicinal chemistry optimization. The compounds demonstrate potent inhibition against Pim-1 and Pim-2 in enzyme assays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service