Skip to Content
Merck
All Photos(1)

Documents

I11704

Sigma-Aldrich

2-Iodotoluene

98%

Synonym(s):

1-Iodo-2-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4I
CAS Number:
Molecular Weight:
218.03
Beilstein:
1634294
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.608 (lit.)

bp

211 °C (lit.)

density

1.713 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1I

InChI

1S/C7H7I/c1-6-4-2-3-5-7(6)8/h2-5H,1H3

InChI key

RINOYHWVBUKAQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven Herbers et al.
Physical chemistry chemical physics : PCCP, 22(20), 11490-11497 (2020-05-12)
The internal rotation of methyl groups and nuclear quadrupole moments of the halogens Cl, Br, I in o-halotoluenes cause complex spectral fine and hyperfine structures in rotational spectra arising from angular momentum coupling. Building on the existing data regarding o-fluorotoluene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service