Key Documents
HAM59000-40
Hamilton® Microlab® 600 instrument syringe
model, 1002 BFP, volume 2.5 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1002 BFP
manufacturer/tradename
Hamilton 59000-40
parameter
10-115 °C temp. range (50-239 °F)
200 psig pressure
I.D. × O.D.
7.29 mm × 10.3 mm
volume
2.5 mL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)