Key Documents
HAM59000-25
Hamilton® Microlab® 600 instrument syringe
model, 1725 BFP, volume 250 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1725 BFP
manufacturer/tradename
Hamilton 59000-25
parameter
10-115 °C temp. range (50-239 °F)
500 psig pressure
I.D. × O.D.
2.30 mm × 7.75 mm
volume
250 μL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)