Skip to Content
Merck
All Photos(2)

Key Documents

H4766

Sigma-Aldrich

Hydrazine hemisulfate salt

≥98%

Synonym(s):

Dihydrazine sulfate, Dihydrazinium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNH2 · 1/2H2SO4
CAS Number:
Molecular Weight:
81.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

SMILES string

NN.NN.OS(O)(=O)=O

InChI

1S/2H4N2.H2O4S/c2*1-2;1-5(2,3)4/h2*1-2H2;(H2,1,2,3,4)

InChI key

DBLJAFVPFHRQSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Hydrogen Generation from Hydrolytic Dehydrogenation of Hydrazine Borane: Discusses the generation of hydrogen from hydrazine hemisulfate and its potential applications in hydrogen energy sources (Tunç et al., 2015).

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You're the flight surgeon: hydrazine exposure.
Jason W Cromar
Aviation, space, and environmental medicine, 84(5), 540-541 (2013-05-30)
Lei Zhou et al.
Methods in molecular biology (Clifton, N.J.), 951, 297-306 (2013-01-09)
Human tear fluid is a complex biological fluid that is actually the extracellular fluid of the surface of the eye and can be considered a dilute protein solution. The quality of the tears are critical for vision as they provide
Michael E Deary et al.
Organic & biomolecular chemistry, 11(2), 309-317 (2012-11-29)
Our recent work has provided new insights into the equilibria and species that exist in aqueous solution at different pHs for the boric acid - hydrogen peroxide system, and the role of these species in oxidation reactions. Most recently, (M.
Yinhu Wang et al.
Biosensors & bioelectronics, 43, 180-185 (2013-01-12)
We report a novel hydrogen peroxide (H2O2) and hydrazine sensor based on low-cost poly(vinylpyrrolidone)-protected silver nanocubes (PVP-AgNCs). The monodisperse silver nanocubes were prepared by adding a trace amount of sodium sulfide in the conventional polyol synthesis for fast reduction of
Tino Wilson Sanchez et al.
Bioorganic & medicinal chemistry, 21(4), 957-963 (2013-01-12)
Human lens epithelium-derived growth factor (LEDGF)/p75 plays an important role in the HIV life cycle by stimulating integrase (IN)-led viral DNA integration into cellular chromosomes. Mechanistic studies show the majority of IN inhibitors chelate magnesium ions in the catalytic active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service