Skip to Content
Merck
All Photos(1)

Key Documents

59767

Sigma-Aldrich

Chlorophenol Red-β-D-galactopyranoside

β-galactosidase, ≥90% (HPLC), powder or crystals

Synonym(s):

CPRG, Chlorophenol Red-β-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22Cl2O10S
CAS Number:
Molecular Weight:
585.41
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Chlorophenol Red-β-D-galactopyranoside, ≥90% (HPLC)

Assay

≥90% (HPLC)

form

powder or crystals

solubility

water: 20 mg/mL, clear, orange to very deep red

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2Cl)C(=C3/C=CC(=O)C(Cl)=C3)\c4ccccc4S(O)(=O)=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C25H22Cl2O10S/c26-15-9-12(5-7-17(15)29)21(14-3-1-2-4-20(14)38(33,34)35)13-6-8-18(16(27)10-13)36-25-24(32)23(31)22(30)19(11-28)37-25/h1-10,19,22-25,28,30-32H,11H2,(H,33,34,35)/b21-12-/t19-,22+,23+,24-,25-/m1/s1

InChI key

YOUWVNCQJOMMEU-AETRGLENSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorophenol red-β-D-galactopyranoside (CPRG) is a long-wavelength yellow dye commonly used for colorimetric assays. It serves as a substrate for β-galactosidase (β-gal) which hydrolyzes CPRG to form chlorophenol red (purple colored). This product is quantified by measuring its absorbance at 570 nm. CPRG is used to identify fecal coliform bacteria. It is about 10 times more sensitive than other chromogenic agents such as o-nitrophenyl-β-galactoside (ONPG). CPRG also has a high rapid turnover.

Application

Chlorophenol Red-β-D-galactopyranoside has been used as a substrate for β-galactosidase (β-gal):
  • in paper-based cell-free protein expression system for target RNA detection
  • in colorimetric detection of multiple clinically relevant mutations using a lacZ output gene
  • in colorimetric assay for bacteria quantification

Other Notes

Measures the expression levels of β-galactosidase in transfected cells. The lacZ gene is a popular reporter gene in transfection reactions because its product, β-galactosidase, is a very stable enzyme.citationcitation
Measures the expression levels of β-galactosidase in transfected cells. The lacZ gene is a popular reporter gene in transfection reactions because its product, β-galactosidase, is a very stable enzyme.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Felgner et al.
The Journal of biological chemistry, 269(4), 2550-2561 (1994-01-28)
The application of cationic liposome reagents has advanced DNA and mRNA transfection research in vitro, and data are accumulating which show their utility for in vivo gene transfer. However, chemical structure-activity data leading to a better mechanistic understanding of their
C Vega et al.
Parasitology research, 95(4), 296-298 (2005-02-01)
We have developed a new pharmacological screening assay for epimastigotes of Trypanosoma cruzi (clone CL-B5) that express the Escherichia coli LacZ gene. The assay is based on determining the activity of the cytoplasmic beta-galactosidase released into the culture on membrane
Generation of antigen-specific, lacZ-inducible T-cell hybrids.
S Malarkannan et al.
Methods in molecular biology (Clifton, N.J.), 156, 265-272 (2000-11-09)
V C Wutor et al.
Chemosphere, 68(4), 622-627 (2007-04-17)
The presence of coliforms in polluted water was determined enzymatically (in situ) by directly monitoring the activity of beta-d-galactosidase (B-GAL) through the hydrolysis of the yellow chromogenic subtrate, chlorophenol red beta-d-galactopyranoside (CPRG), which produced a red chlorophenol red (CPR) product.
D C Eustice et al.
BioTechniques, 11(6), 739-740 (1991-12-01)
A sensitive method has been developed for the detection of E. coli beta-galactosidase in transfected HeLa cells. The chromogenic substrate, CPRG (chlorophenol red-beta-D-galactopyranoside), was compared with ONPG (o-nitrophenyl-beta-D-galactopyranoside) by kinetic analysis with purified beta-galactosidase. The Km for CPRG was 1.35

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service