Skip to Content
Merck
All Photos(2)

Documents

GF48969905

Ytterbium

lump, 25g, max. size 25mm, 99.9%

Synonym(s):

Ytterbium, YB006100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12141616
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 489-699-05

resistivity

28 μΩ-cm, 20°C

max. lump size

25 mm

bp

1196 °C (lit.)

mp

824 °C (lit.)

density

6.54 g/mL at 25 °C (lit.)

SMILES string

[Yb]

InChI

1S/Yb

InChI key

NAWDYIZEMPQZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ru Zhou et al.
Journal of nanoscience and nanotechnology, 14(6), 4196-4200 (2014-04-18)
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission
Wanyue Shao et al.
Journal of nanoscience and nanotechnology, 14(5), 3887-3892 (2014-04-17)
Water-soluble BaYF5:Yb3+/Er3+ nanoparticles with the surface functionalized by a layer of poly(acrylic acid) (PAA) were synthesized via a facile one-step PAA-assisted hydrothermal method. Bovine serum albumin (BSA) protein was conjugated with BaYF5:Yb3+/Er3+ upconversion nanoparticles via free carboxylic acid groups on
Sethuraman Gayathri et al.
Carbohydrate polymers, 121, 302-308 (2015-02-11)
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region.
Liming Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4710-4713 (2014-04-18)
LiYbF4:Yb, Er nanoparticles have been successfully synthesized by thermal decomposition of multiple trifluoroacetic acid salts. The SEM and TEM results show the size of the LiYF4:Yb, Er nanoparticles is about 100 nm in diagonal line, and the morphology of the
Li Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4313-4319 (2014-04-18)
Lutetium oxide nanocrystals codoped with Tm3+ and Yb3+ have been successfully synthesized via adjusting the pH values of the precursor solution in a hydrothermal method followed by a subsequent calcination process. The samples were systematically characterized by X-ray diffraction, field-emission

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service