GF21238332
Ytterbium
lump, 50g, max. size 25mm, 99.9%
Synonym(s):
Ytterbium, YB006100
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12141616
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.90%
form
tubes
manufacturer/tradename
Goodfellow 212-383-32
resistivity
28 μΩ-cm, 20°C
max. lump size
25 mm
bp
1196 °C (lit.)
mp
824 °C (lit.)
density
6.54 g/mL at 25 °C (lit.)
SMILES string
[Yb]
InChI
1S/Yb
InChI key
NAWDYIZEMPQZHO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sethuraman Gayathri et al.
Carbohydrate polymers, 121, 302-308 (2015-02-11)
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region.
Ying Xie et al.
Journal of nanoscience and nanotechnology, 14(6), 4486-4489 (2014-04-18)
Eu3+ and Yb3+ codoped Y2O3 phosphors were synthesized by the sol-gel method. The phosphors possess absorption in the region of 300-550 nm, exhibiting an intense NIR emission of Yb3+ around 1000 nm, which is suitable for matching the maximum spectral
Pr3+/Yb3+ co-doped beta-phase NaYF4 microprisms: controlled synthesis and upconversion luminescence.
Wei Gao et al.
Journal of nanoscience and nanotechnology, 14(6), 4308-4312 (2014-04-18)
Pr3+/Yb3+ co-doped hexagonal NaYF4(beta-NaYF4) microprisms were synthesized by the hydrothermal method, and ethylenediaminetetraacetic acid (EDTA) was introduced to control the size of the microcrystal samples. Bright upconverted fluorescence emission was observed when the samples were excited with an infrared (IR)
Liming Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4710-4713 (2014-04-18)
LiYbF4:Yb, Er nanoparticles have been successfully synthesized by thermal decomposition of multiple trifluoroacetic acid salts. The SEM and TEM results show the size of the LiYF4:Yb, Er nanoparticles is about 100 nm in diagonal line, and the morphology of the
Ru Zhou et al.
Journal of nanoscience and nanotechnology, 14(6), 4196-4200 (2014-04-18)
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service