Skip to Content
Merck
All Photos(2)

Key Documents

GF23443646

Magnesium

wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%

Synonym(s):

Magnesium, MG005120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
MDL number:
UNSPSC Code:
12141502
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 621 °C)

Assay

99.90%

form

wire

autoignition temp.

950 °F

manufacturer/tradename

Goodfellow 234-436-46

resistivity

4.46 μΩ-cm, 20°C

L × diam.

0.5 mm × 0.4 mm

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Yang
Biochemistry, 53(17), 2793-2803 (2014-04-11)
Y-Family DNA polymerases specialize in translesion synthesis, bypassing damaged bases that would otherwise block the normal progression of replication forks. Y-Family polymerases have unique structural features that allow them to bind damaged DNA and use a modified template base to
Charles L Loprinzi et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(10), 997-1005 (2013-12-04)
Cumulative neurotoxicity is a prominent toxicity of oxaliplatin-based therapy. Intravenous calcium and magnesium have been extensively used to reduce oxaliplatin-induced neurotoxicity. This trial was designed to definitively test whether calcium/magnesium decreases oxaliplatin-related neurotoxicity. In all, 353 patients with colon cancer
Jeroen H F de Baaij et al.
Physiological reviews, 95(1), 1-46 (2014-12-30)
Magnesium (Mg(2+)) is an essential ion to the human body, playing an instrumental role in supporting and sustaining health and life. As the second most abundant intracellular cation after potassium, it is involved in over 600 enzymatic reactions including energy
Jeremy R Keown et al.
The Biochemical journal, 464(3), 413-423 (2014-09-24)
Most plants contain two isoforms of ribulose-1,5-bisphosphate carboxylase/oxygenase (Rubisco) activase (Rca), a chloroplast protein that maintains the activity of Rubisco during photosynthesis. The longer (α-) Rca isoform has previously been shown to regulate the activity of Rubisco in response to
Parisa Asghari et al.
Circulation research, 115(2), 252-262 (2014-05-03)
Single-tilt tomograms of the dyads in rat ventricular myocytes indicated that type 2 ryanodine receptors (RYR2s) were not positioned in a well-ordered array. Furthermore, the orientation and packing strategy of purified type 1 ryanodine receptors in lipid bilayers is determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service