Skip to Content
Merck
All Photos(1)

Key Documents

13103

Sigma-Aldrich

Magnesium

ribbon, ≥99.0% Mg basis

Synonym(s):

Magnesium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

Assay

≥99.0% Mg basis

form

ribbon

autoignition temp.

950 °F

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

size

~3 mm × 0.2 mm

impurities

≤0.05% insoluble in HCl

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

cation traces

Cu: ≤50 mg/kg
Fe: ≤200 mg/kg
Pb: ≤100 mg/kg

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium ribbons can be used to prepare Grignard reagent to synthesize a wide range of reaction intermediates.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microwave-induced electrostatic etching: generation of highly reactive magnesium for application in Grignard reagent formation.
van D K B H, et al.
Organic & Biomolecular Chemistry, 8(7), 1688-1694 (2010)
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Benjamin Chaigne-Delalande et al.
Science (New York, N.Y.), 341(6142), 186-191 (2013-07-13)
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free
Jochen Halfar et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(49), 19737-19741 (2013-11-20)
Northern Hemisphere sea ice has been declining sharply over the past decades and 2012 exhibited the lowest Arctic summer sea-ice cover in historic times. Whereas ongoing changes are closely monitored through satellite observations, we have only limited data of past
Pallavi K Choudhary et al.
RNA (New York, N.Y.), 20(1), 36-45 (2013-11-19)
Mg(2+) has been shown to modulate the function of riboswitches by facilitating the ligand-riboswitch interactions. The btuB riboswitch from Escherichia coli undergoes a conformational change upon binding to its ligand, coenzyme B12 (adenosyl-cobalamine, AdoCbl), and down-regulates the expression of the

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service