Skip to Content
Merck
All Photos(1)

Key Documents

G2354

Sigma-Aldrich

α-D(+)-Glucose pentaacetate

99%

Synonym(s):

1,2,3,4,6-Penta-O-acetyl-α-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
Beilstein:
98852
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D ≥+98°, c = 1 in ethanol

mp

109-111 °C (lit.)

SMILES string

CC(=O)OC[C@H]1O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3/t12-,13-,14+,15-,16+/m1/s1

InChI key

LPTITAGPBXDDGR-LJIZCISZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-D(+)-Glucose pentaacetate also known as 1,2,3,4,6-penta-O-acetyl-α-D-glucopyranose, is an acetylated sugar that has wide applications in organic synthesis.

Application

α-D(+)-Glucose pentaacetate is used as a model compound to study the stereochemistry of carbohydrates through spectroscopic techniques such as Vibrational circular dichroism (VCD). Additionally, it has been used as a standard in the analysis of monosaccharide and polysaccharide components by gas chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of polysaccharides and monosaccharides in the root mucilage of maize (Zea mays L.) by gas chromatography
HI Osborn
Journal of Chromatography A, 831, 267-276 (1999)
Acetate groups as probes of the stereochemistry of carbohydrates: a vibrational circular dichroism study
PK Bose
Carbohydrate Research, 322, 135-141 (1999)
Monosaccharide esters: new tools in biomedical research.
W J Malaisse
Molecular genetics and metabolism, 65(2), 129-142 (1998-10-27)
W J Malaisse et al.
Archives of biochemistry and biophysics, 381(1), 61-66 (2000-10-06)
Hepatocytes from fed rats were incubated for 120 min in the presence of alpha-D-[1,2-13C]glucose pentaacetate (1.7 mM), both D-[1,2-13C]glucose (1.7 mM) and acetate (8.5 mM), alpha-D-glucose penta[2-13C]acetate (1.7 mM), or D-[1,2-13C]glucose (8.3 mM). The amounts of 13C-enriched L-lactate and D-glucose
W J Malaisse et al.
The American journal of physiology, 275(6 Pt 1), E993-E1006 (1998-12-09)
The metabolism of beta-L-glucose pentaacetate and its interference with the catabolism of L-[U-14C]glutamine, [U-14C]palmitate, D-[U-14C]glucose, and D-[5-3H]glucose were examined in rat pancreatic islets. Likewise, attention was paid to the effects of this ester on the biosynthesis of islet peptides, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service