Skip to Content
Merck
All Photos(2)

Key Documents

F203

Sigma-Aldrich

Farnesol

95%

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
222.37
Beilstein:
1763926
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.489 (lit.)

bp

149 °C/4 mmHg (lit.)

density

0.886 g/mL at 20 °C (lit.)

SMILES string

C/C(CC/C=C(C)/CCC=C(C)C)=C\CO

InChI

1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+

InChI key

CRDAMVZIKSXKFV-YFVJMOTDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gisselle Moraima Chávez-Andrade et al.
Archives of oral biology, 107, 104481-104481 (2019-07-25)
This study aimed to evaluate the antimicrobial and biofilm anti-adhesion activities of poly(vinyl alcohol)-coated silver nanoparticles (AgNPs-PVA) and farnesol against Enterococcus faecalis, Candida albicans or Pseudomonas aeruginosa. Minimum inhibitory concentration (MIC) and minimum microbicidal concentration (MMC) of the solutions, as
Efrat Mashiach-Farkash et al.
Oncotarget, 3(6), 629-639 (2012-07-11)
Neurofibromin regulates cell motility via three distinct GTPase pathways acting through two different domains, the Ras GTPase-activating protein-related domain (GRD) and the pre-GRD domain. First, the GRD domain inhibits Ras-dependent changes in cell motility through the mitogen activated protein cascade.
N Charette et al.
Cell death & disease, 4, e471-e471 (2013-01-26)
Ras activation is a frequent event in human hepatocarcinoma that may contribute to resistance towards apoptosis. Salirasib is a ras and mTOR inhibitor that induces a pro-apoptotic phenotype in human hepatocarcinoma cell lines. In this work, we evaluate whether salirasib
Najla Nasr et al.
Blood, 120(4), 778-788 (2012-06-09)
Macrophages are key target cells for HIV-1. HIV-1(BaL) induced a subset of interferon-stimulated genes in monocyte-derived macrophages (MDMs), which differed from that in monocyte-derived dendritic cells and CD4 T cells, without inducing any interferons. Inhibition of type I interferon induction
Kit Kennedy et al.
PLoS biology, 17(7), e3000376-e3000376 (2019-07-19)
Apicomplexan parasites possess a plastid organelle called the apicoplast. Inhibitors that selectively target apicoplast housekeeping functions, including DNA replication and protein translation, are lethal for the parasite, and several (doxycycline, clindamycin, and azithromycin) are in clinical use as antimalarials. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service